Skip to Content
Merck
All Photos(1)

Key Documents

STREAMTUB

Millipore

Silicone Tubing

silicon tube, for use with EZ-Fit® Filtration Manifold, for use with Membrane filtration system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

silicon tube

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Stream®

L

5 m

application(s)

bioburden testing
food and beverages
microbiology
pharmaceutical
water monitoring

compatibility

for use with EZ-Fit® Filtration Manifold
for use with Membrane filtration system

shipped in

ambient

General description

The silicone tubing is designed to be used with all filtration systems as an external drain tube. The tube is used to connect the EZ-Stream® Pump to the manifold or filtration apparatus, and the discard tubing is placed to drain. The tubing is autoclavable, 5 m, long with an internal diameter of 9.5 mm.

Application

The silicone tubing is designed to be used with all membrane filtration systems as an external drain tube for general microbial analysis of beer, bottled water, cider, soft drink, sports drink, wine, dairy, and environmental analysis.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-STREAM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service