EZFITSAM3
EZ-Fit® Manifold
3-place manifold, for use with EZ-Fit® Filtration Unit, stainless steel
Synonym(s):
EZ-Fit® Filtration Manifold, EZ-Fit® manifold for EZ-Stream® vacuum pump, high throughput application, Filtration Manifold Base, Manifold Base, Manifold head
About This Item
Recommended Products
material
stainless steel
Sterilization Compatibility
autoclavable compatible
feature
3-place manifold
packaging
pkg of 1 unit
manufacturer/tradename
EZ-Fit®
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
for use with EZ-Fit® Filtration Unit
shipped in
ambient
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The EZ-Fit® Filtration Manifold Base can accommodate three filtration heads at a time, which would mean three samples can be tested at one go, and the ergonomic design enables operation under laminar flow hood and save laboratory bench space.
The 2-way valve system of the filtration manifold prevents the user to accidentally touch the filtration head while opening or closing the valve.
Application
Features and Benefits
- Different filtration heads fit both reusable and disposable filtration devices.
- Each component is removable for deeper cleaning.
- Autoclavable parts.
- Quick fit connections for vacuum tubing.
- Low height for easier use inside Laminar Flow Hood.
Packaging
EZFITMHTA3 x1
Analysis Note
Legal Information
accessory
configured for
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
The EZ Product Family optimizes your microbial testing workflow to deliver reliable and reproducible microbial enumeration test results. The product lineup consists of filtration devices, manifolds, calibrated vacuum sources, and rapid microbial detection platforms.
The EZ Product Family optimizes your microbial testing workflow to deliver reliable and reproducible microbial enumeration test results. The product lineup consists of filtration devices, manifolds, calibrated vacuum sources, and rapid microbial detection platforms.
The EZ Product Family optimizes your microbial testing workflow to deliver reliable and reproducible microbial enumeration test results. The product lineup consists of filtration devices, manifolds, calibrated vacuum sources, and rapid microbial detection platforms.
The EZ Product Family optimizes your microbial testing workflow to deliver reliable and reproducible microbial enumeration test results. The product lineup consists of filtration devices, manifolds, calibrated vacuum sources, and rapid microbial detection platforms.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service