Skip to Content
Merck
All Photos(3)

Key Documents

STBMTSB12

Millipore

Tryptic Soy Broth Ready-to-use Bottles

bottle capacity 125 mL, bottle filling volume 100 mL, closure type, green screw cap with septum, pack of 12 bottles

Synonym(s):

Caso bouillon, Soybean Casein Digest Broth, TSB, Trypcase soya broth, Tryptic soy broth, Tryptone soya broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
eCl@ss:
32014001
NACRES:
NA.74

Agency

in accordance with EP 2.6.1
in accordance with JP 4.06
in accordance with USP 71

Quality Level

description

Bottle with screw cap and septum, For the detection of aerobic bacteria and fungi

sterility

sterile; autoclaved

form

liquid

shelf life

12 mo.

feature

closure type green screw cap with septum
ready-to-use

packaging

pack of 12 bottles

bottle capacity

125 mL

bottle filling volume

100 mL

capacity

125 mL

input

sample type pharmaceutical(s)

pH

7.3

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest®

shipped in

ambient

storage temp.

2-30°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Soybean Casein Digest Broth is a sterility medium and a critical component of our STERITEST® solution. Composition and growth performance of this medium comply with the European Pharmacopeia (EP) 2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06. Our manufacturing approach ensures the highest level of quality for our media, improving accuracy and significantly reducing the risk of incorrect interpretation and false results. The appearance of the media is a light-yellow clear liquid. It has a pH of 7.3 ± 0.2 and the storage temperature is 2-25°C. The incubation temperature varies between 20-25°C, with an incubation time of 7-14 days. One box of product contains 12 bottles of 100ml of Medium. Each bottle has a screw cap with a septum, which minimizes the risks of cross-contamination and optimizes the disinfection procedures.

Application

Soybean-Casein Digest Medium is intended for the detection of aerobic bacteria and fungi. The QC organisms include Bacillus subtilis (ATCC 6633), Candida albicans (ATCC 10231), Aspergillus brasiliensis (niger) (ATCC 16404), Staphylococcus aureus (ATCC 6538) and Pseudomonas aeruginosa (ATCC 9027). This medium is used for sterility testing by membrane filtration or direct inoculation. It is also used as a pre-enrichment broth for nonsterile products.
Sterility testing (membrane filtration and direct inoculation)
Bioburden testing

Components

Screw cap with septum

Specifications

Incubation Temperature: 20–25 °C
Incubation Time: 7–14 days
United-States Pharmacopeia, European Pharmacopeia, Japanese Pharmacopeia

Culture Medium

Trypticase Soy

Analysis Note

Turbidity indicates growth

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

Sterile Isopropyl Myristate (IPM) enhances filterability for sterility testing of viscous products using the Steritest® NEO device.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service