Skip to Content
Merck
All Photos(3)

Key Documents

STBMRFA12DP

Millipore

Fluid A - Ready-to-use Rinse Fluid

bottle capacity 125 mL, bottle filling volume 100 mL, closure type, Black screw cap with septum and protector, double packed of 12 bottles

Synonym(s):

Fluid A, Double packed, Rinse Fluid, Washing Fluid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214
eCl@ss:
32014001
NACRES:
NA.74
Pricing and availability is not currently available.

Agency

in accordance with EP 2.6.1
in accordance with JP 4.06
in accordance with USP 71

Quality Level

sterility

sterile; ethylene oxide treated

form

liquid

feature

closure type Black screw cap with septum and protector
ready-to-use

packaging

double packed of 12 bottles

bottle capacity

125 mL

bottle filling volume

100 mL

capacity

125 mL

input

sample type pharmaceutical(s)

pH

7.1

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest®

shipped in

ambient

storage temp.

2-30°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Fluid A is a general rinse buffer in compliance with the European Pharmacopeia (EP) 2.6.1, United States Pharmacopeia (USP) 71 and Japanese Pharmacopeia (JP) 4.06. The sterilized double Tyvek® packaging increases the test method reliability, allowing to streamline the cleaning procedures, minimize the risk of cross-contamination in laminar flow hoods and secure an efficient decontamination of isolator chambers. The product is a clear colorless liquid with a pH of 7.1 ± 0.2 at 25°C. It can be stored at 2-25°C (protected from light) for 12 months. A single box contains 12 bottles of 100ml of fluid, each bottle with a screw cap with septum closure that reduces the risk of cross-contamination and optimizes the disinfection procedures.

Application

Fluid A is used to rinse aqueous solutions during sterility testing by membrane filtration. It is also used to dilute soluble solids for the same application. This product is recommended as a rinsing fluid for membrane filtration of non-sterile products. It is excellent for dissolving or diluting samples, reconstituting commercial microorganisms, or as a transport medium for microorganisms.
Sterility testing
Bioburden testing

Components

Screw cap with septum

Culture Medium

Fluid A

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company

recommended

Product No.
Description
Pricing

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service