Skip to Content
Merck
All Photos(3)

Key Documents

STBMFTM12DP

Millipore

Fluid Thioglycollate Medium, Double packed

bottle capacity 125 mL, bottle filling volume 100 mL, closure type, Red screw cap with septum and protector, double packed of 12 bottles

Synonym(s):

Fluid Thioglycollate Media, Fluid Thioglycollate Medium (FTGM), Thioglycolate broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106225
eCl@ss:
32014001
NACRES:
NA.74

Agency

in accordance with EP 2.6.1
in accordance with JP 4.06
in accordance with USP 71

Quality Level

sterility

sterile; ethylene oxide treated

form

liquid

feature

closure type Red screw cap with septum and protector
ready-to-use

packaging

double packed of 12 bottles

bottle capacity

125 mL

bottle filling volume

100 mL

input

sample type pharmaceutical(s)

pH

7.1

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest®

shipped in

ambient

storage temp.

2-30°C

suitability

anaerobic bacteria

General description

Fluid Thioglycollate Medium (FTM) is a critical component of our STERITEST® solution. The product has been formulated and tested to meet the requirements of the EP 2.6.1, USP 71, and JP 4.06. Our manufacturing approach ensures the highest level of quality for our media, improves accuracy and significantly reduces the risk of incorrect interpretation and false results. The sterilized double Tyvek® packaging allows to streamline the cleaning procedures, to minimize the risk of cross-contamination in laminar flow hoods and to secure an efficient decontamination of isolator chambers. The fluid appears light yellow, slightly opalescent and viscous with a pink ring in suspension ≤ 1 cm. The pH of the product is 7.1 ± 0.2 and can be stored at 2-25°C. The incubation temperature varies between 30-35°C, with an incubation time of 7-14 days. The product is supplied as 12 bottles of 100 mL of medium, screw cap with septum closure that reduces the risk of cross-contamination and growth inhibition.

Application

Fluid Thioglycollate Medium (FTM) is primarily used for the detection of anaerobic bacteria. However, it also enables aerobic bacterial detection. The QC organisms include Clostridium sporogenes (ATCC 19404), Staphylococcus aureus (ATCC 6538) and Pseudomonas aeruginosa (ATCC 9027). This medium is used for sterility testing by membrane filtration or direct inoculation.
Sterility testing (membrane filtration and direct inoculation)
Bioburden testing

Components

Screw cap with septum

Culture Medium

Fluid Thioglycollate

Analysis Note

Turbidity indicates growth

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Sterility testing media options facilitate direct inoculation for non-filterable samples, including soybean casein digest and Fluid Thioglycollate medium.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service