MX0819
tert-Butyl methyl ether
Meets ACS Specifications GR ACS
Synonym(s):
Methyl-t-Butyl Ether, tert-Butyl Methyl Ether,MTBE, MTBE
About This Item
Recommended Products
Product Name
tert-Butyl methyl ether, Meets ACS Specifications GR ACS
vapor pressure
268 hPa ( 20 °C)
Quality Level
Assay
≥99% (GC)
form
liquid
impurities
≤0.05% Water (H2O)
≤1 ppm Peroxide (as H2O2)
evapn. residue
≤0.001%
color
APHA: ≤10
refractive index
n25/D 1.365-1.367
bp
55.3 °C/1013 hPa
mp
-108.6 °C
transition temp
flash point -28 °C
solubility
42 g/L
density
0.735-0.738 g/mL at 25 °C
shipped in
ambient
storage temp.
room temp
SMILES string
O(C(C)(C)C)C
InChI
1S/C5H12O/c1-5(2,3)6-4/h1-4H3
InChI key
BZLVMXJERCGZMT-UHFFFAOYSA-N
Related Categories
Application
Analysis Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-18.4 °F - closed cup
Flash Point(C)
-28 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service