1.01849
tert-Butyl methyl ether
for analysis EMSURE® ACS
Synonym(s):
tert-Butyl methyl ether, Methyl tert-butyl ether, MTB, MTBE
About This Item
grade
ACS reagent
Quality Level
vapor pressure
268 hPa ( 20 °C)
product line
EMSURE®
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
460 °C
potency
>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)
expl. lim.
1.5-8.5 % (v/v)
impurities
≤0.000002% B (Boron)
≤0.000002% Co (Cobalt)
≤0.000005% Cd (Cadmium)
≤0.00001% Ba (Barium)
≤0.00005% Al (Aluminium)
≤0.00005% Ca (Calcium)
≤0.0001% Peroxide (as H2O2)
≤0.0005 meq/g Acidity
≤0.03% Water
evapn. residue
≤0.001%
color
APHA: ≤10
kinematic viscosity
0.464 cSt(20 °C)
bp
55.3 °C/1013 hPa
mp
-108.6 °C
transition temp
flash point -28 °C
density
0.74 g/cm3 at 20 °C
cation traces
Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%
storage temp.
2-30°C
SMILES string
O(C(C)(C)C)C
InChI
1S/C5H12O/c1-5(2,3)6-4/h1-4H3
InChI key
BZLVMXJERCGZMT-UHFFFAOYSA-N
General description
Application
- Application of liquid chromatography coupled to data-independent acquisition mass spectrometry for the metabolic profiling of N-ethyl heptedrone.: This study utilized liquid chromatography-mass spectrometry to perform metabolic profiling, highlighting the advanced analytical capabilities of tert-Butyl methyl ether in complex sample analysis. The research demonstrated its efficacy in detecting various metabolites, contributing to the development of high-throughput analytical techniques (Mazzarino et al., 2021).
- Extraction of Lipids from Liquid Biological Samples for High-Throughput Lipidomics.: This paper explored the use of tert-Butyl methyl ether for lipid extraction from biological samples, facilitating high-throughput lipidomic analyses. The method′s efficiency and reliability were demonstrated, making it valuable for large-scale biochemical studies (Furse et al., 2020).
- A sensitive and efficient procedure for the high-throughput determination of nine urinary metabolites of pyrethroids by GC-MS/MS and its application in a sample of Japanese children.: This research presented a high-throughput method for analyzing urinary metabolites using GC-MS/MS, where tert-Butyl methyl ether played a crucial role in sample preparation. The study emphasized the method′s sensitivity and applicability in large-scale population studies (Ueda et al., 2018).
- Determination of ochratoxin A in baby food by immunoaffinity column cleanup with liquid chromatography: interlaboratory study.: This study employed tert-Butyl methyl ether in the cleanup process for ochratoxin A detection in baby food, demonstrating its utility in ensuring food safety through robust analytical techniques (Burdaspal et al., 2001).
Other Notes
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-18.4 °F - closed cup
Flash Point(C)
-28 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service