MHA000P2S
m-Heterotrophic Plate Count Broth
2ml ampoule for the recovery of stressed heterotrophic bacteria found in various types of water. Also suitable for water samples with low counts.
Synonym(s):
m-HPC Broth
About This Item
Recommended Products
Quality Level
sterility
sterile
form
liquid
shelf life
12 mo.
feature
ready-to-use
packaging
pack of 50 tests
technique(s)
cell counting: suitable (total viable count (TVC))
capacity
2 mL
filling volume
2 mL
color
clear tan to light amber
input
sample type cider
sample type beverage(s)
sample type cosmetics
sample type bottled water
sample type food(s)
sample type beer
sample type wine
sample type soft drink
sample type sport drink
pH
7.1±0.2 (25.0 °C)
application(s)
clinical
cosmetics
environmental
food and beverages
industrial qc
microbiology
water monitoring
compatibility
for use with MF-method
for use with Microfil®
for use with MicropreSure®
for use with Milliflex®
shipped in
wet ice
storage temp.
2-8°C
suitability
heterotrophic bacteria
Related Categories
General description
Application
Packaging
Specifications
Preparation Note
Culture Medium
Analysis Note
E. coli (ATCC 25922)
S. epidermidis (ATCC 14990)
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service