MXPESPTCL
MicropreSure® Sanitary Sampling Valve
1-1/2" Tri-clover valve for On-line sampling and filtration of aqueous liquids
Synonym(s):
Stainless steel sampling valve
About This Item
Recommended Products
material
316L stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
MicropreSURE®
valve size
1-1/2 in.
application(s)
bioburden testing
food and beverages
pharmaceutical
water monitoring
compatibility
1-1/2" Tri-clover valve for On-line sampling and filtration of aqueous liquids
shipped in
ambient
Related Categories
General description
Application
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
MicropreSure® on-line filtration system for in-process microbiology testing
MicropreSure® on-line filtration system for in-process microbiology testing
MicropreSure® on-line filtration system for in-process microbiology testing
MicropreSure® on-line filtration system for in-process microbiology testing
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service