Skip to Content
Merck
All Photos(1)

Key Documents

MAB2019Z

Sigma-Aldrich

Anti-Integrin αVβ5 Antibody, clone 15F11, Azide Free

clone 15F11, Chemicon®, from mouse

Synonym(s):

MAB2019

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

15F11, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

Specificity

Reactive with human integrin alphaVbeta5. Does not recognize integrin alphaVbeta3 or alpha5beta1. The antibody binds to the alphaVbeta5 complex. The monoclonal antibody MAB2019Z has a Kd of 2.8 nM in Mn2+, 4.0 nM in Ca2+and 2.9 nM in buffer lacking cation. Thus, cations do not significantly affect the binding of the antibody to alphaVbeta5. Monoclonal antibody MAB2019Z does not block ligand binding to purified alphaVbeta5, nor does it block cell adhesion mediated through this receptor.

Immunogen

Generated against human integrin alphaVbeta5 purified from placental tissue and depleted of integrin alphaVbeta3.

Application

Anti-Integrin αVβ5 Antibody, clone 15F11, Azide Free is an antibody against Integrin αVβ5 for use in ELISA, FC, IP & IC.
Immunoprecipitation

Flow Cytometry: MG-63 cells have been harvested, blocked for 20 minutes at 4C in 20% normal human serum then washing in medium containing 10% FBS. 5X10E5 cells per tube were subsequently incubated with primary antibody for 30 min on ice, washed with medium, and incubated with fluorescent-conjugated secondary antibody.

Immunocytochemistry: 15μg/mL dilution for immunostaining of cells adhering to vitronectin-coated coverslips and fixed in 1% paraformaldehyde containing 0.5% Triton X-100 for 20 minutes. Incubation with MAB2019Z antibody for 30 minutes at 22C, followed by detection with a fluorescein-conjugated secondary antibody.

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Purified immunoglobulin. Liquid at 1 mg/mL in PBS, containing no preservatives.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stimulation of glioma cell motility by expression, proteolysis, and release of the L1 neural cell recognition molecule.
Yang, M; Adla, S; Temburni, MK; Patel, VP; Lagow, EL; Brady, OA; Tian, J; Boulos, MI; Galileo, DS
Cancer Cell International null
Khine Kyaw Oo et al.
BMC cancer, 21(1), 65-65 (2021-01-16)
Chemoresistance is one of the main problems in treatment of cancer. Periostin (PN) is a stromal protein which is mostly secreted from cancer associated fibroblasts in the tumor microenvironment and can promote cancer progression including cell survival, metastasis, and chemoresistance.
Edith Charlier et al.
Bone research, 8, 35-35 (2020-10-22)
We previously reported 18FPRGD2 uptake by the coxofemoral lining, intervertebral discs and facet joint osteophytes in OA using PET/SCAN imaging. However, the molecular mechanism by which the PRGD2 tracer interacts with joint tissues and osteophytes in OA remains unclear. As
Aliakbar Taherian et al.
BMC cancer, 11, 293-293 (2011-07-15)
Integrins are used as prognostic indicators in breast cancer. Following engagement with extracellular matrix proteins, their signaling influences numerous cellular processes including migration, proliferation, and death. Integrin signaling varies between cell types through differential expression of integrin subunits, and changes
Daniel Vial et al.
Journal of cell science, 121(Pt 10), 1661-1670 (2008-05-01)
The plasminogen activation system regulates matrix remodeling through both proteolytic and non-proteolytic mechanisms. Studies were undertaken to determine the effects of the plasminogen activator inhibitor 1 (PAI1) on the assembly of the fibronectin matrix. The addition of PAI1 to MG-63

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service