Skip to Content
Merck
All Photos(1)

Key Documents

MAB1961Z

Sigma-Aldrich

Anti-Integrin αVβ5 Antibody, clone P1F6, azide free

clone P1F6, Chemicon®, from mouse

Synonym(s):

MAB1961

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

P1F6, monoclonal

species reactivity

chicken, human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGAV(3685)

General description

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha V chain. Alpha chain V undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.

Specificity

Reacts with the human integrin alphaVbeta5.

Immunogen

Human UCLA P3 cells (lung carcinoma)

Application

Detect Integrin αVβ5 using this Anti-Integrin αVβ5 Antibody, clone P1F6, azide free validated for use in FC, IH.
Inhibition of attachment of HT29 cells to vitronectin: >1:1,000

Inhibition of attachment of carcinoma and melanoma cells to vitronectin

Immunohistochemistry: for use on acetone or paraformaldehyde fixed tissue. (Staining of formalin-fixed tissues REQUIRES boiling tissue sections in 1mM EDTA, pH 8.0, for 10-20 min followed by cooling at RT for 20 min.)

Immunocytochemistry: for use on human carcinomas, normal epithelium and keratinocytes 1:1500 (Acetone Fixed Only)

Immunoprecipitation

FACS

EIA

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Integrins

Physical form

Format: Purified
Protein A purified
Purified immunoglobulin in 0.01M phosphate buffer, pH 7.1, 0.15M NaCl containing no preservatives.

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Epithelium, keratinocytes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leah E Cole et al.
Infection and immunity, 70(11), 6158-6165 (2002-10-16)
Haemophilus ducreyi is the etiologic agent of the sexually transmitted genital ulcer disease chancroid. H. ducreyi serum resistance protein A (DsrA) is a member of a family of multifunctional outer membrane proteins that are involved in resistance to killing by
M Chen et al.
Cell death and differentiation, 21(5), 797-810 (2014-01-28)
Isthmin (ISM) is a secreted 60-kDa protein that potently induces endothelial cell (EC) apoptosis. It suppresses tumor growth and angiogenesis in mice when stably overexpressed in cancer cells. Although αvβ5 integrin serves as a low-affinity receptor for ISM, the mechanism
Adam M Sonabend et al.
Stem cells (Dayton, Ohio), 26(3), 831-841 (2008-01-15)
Gene therapy represents a promising treatment alternative for patients with malignant gliomas. Nevertheless, in the setting of these highly infiltrative tumors, transgene delivery remains a challenge. Indeed, viral vehicles tested in clinical trials often target only those tumor cells that
ADAM-9 (MDC-9/meltrin-gamma), a member of the a disintegrin and metalloproteinase family, regulates myeloma-cell-induced interleukin-6 production in osteoblasts by direct interaction with the alpha(v)beta5 integrin.
Karadag, A; Zhou, M; Croucher, PI
Blood null
Anti-integrin monoclonal antibody CNTO 95 enhances the therapeutic efficacy of fractionated radiation therapy in vivo.
Ning, S; Nemeth, JA; Hanson, RL; Forsythe, K; Knox, SJ
Molecular Cancer Therapeutics null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service