Skip to Content
Merck
All Photos(1)

Key Documents

MAB1120

Sigma-Aldrich

Anti-Insulin-like Growth Factor-1 Receptor Antibody, α-subunit, clone 24-31

clone 24-31, Chemicon®, from mouse

Synonym(s):

IGF-IR, CD221

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

24-31, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IGF1R(3480)

Specificity

Recognizes the alpha-subunit of type I Insulin-like Growth Factor Receptor (IGF-1R). It shows no cross-reaction with insulin receptor. Its epitope localizes between a.a. 283-440 (exon 4-6) of IGF-1R. Both IGF-1R and insulin receptor are synthesized as a single polypeptide which is glycosylated and proteolytically cleaved to give the alpha- and beta-subunits, which are disulfide-linked in a beta-alpha-alpha-beta configuration in the mature receptor. The a-subunit is completely extracellular, while the beta-subunit spans the membrane and the intracellular portion has intrinsic tyrosine kinase activity. It is assumed that, as for the insulin receptor, the tyrosine kinase activity is essential for IGF-1 action.

Weakly reacts with Rabbit.

Immunogen

IGF-1R/3T3 mouse fibroblasts transfected with human type I IGF-receptor cDNA.

Application

Acts as a Weak IGF-like Agonist in Bioassays (Thymidine incorporation into DNA)

Immunoprecipitation (Use Protein G): 2μg antibody/mg protein lysate.

Immunohistochemistry (Frozen and formalin/paraffin): Use antibody at 2-4ug/mL for 30 min. at room temperature. Staining of formalin-fixed tissues requires boiling tissue sections in 1 mM EDTA, pH 8.0, for 10-20 min. followed by cooling at room temperature for 20 minutes. Please note that EDTA is better than citrate for epitope unmasking.

Optimal working dilutions must be determined by end user.
Anti-Insulin-like Growth Factor-1 Receptor Antibody, α-subunit, clone 24-31 detects level of Insulin-like Growth Factor-1 Receptor & has been published & validated for use in IH(P) & IP.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Physical form

Format: Purified
Purified from ascites fluid by Protein G chromatography. Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Articles of the Hungarian Medical Journal from September, 2006].
Orvosi hetilap, 147(35-38), 1673-1864 (2006-09-03)
Amy M Smith et al.
Frontiers in cellular neuroscience, 16, 1047928-1047928 (2022-11-26)
The study of microglia isolated from adult human brain tissue provides unique insight into the physiology of these brain immune cells and their role in adult human brain disorders. Reports of microglia in post-mortem adult human brain tissue show regional
Phase I/II trial and pharmacokinetic study of cixutumumab in pediatric patients with refractory solid tumors and Ewing sarcoma: a report from the Children's Oncology Group.
Malempati, S; Weigel, B; Ingle, AM; Ahern, CH; Carroll, JM; Roberts, CT; Reid et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology null
Becky K Brisson et al.
PloS one, 7(9), e45588-e45588 (2012-10-03)
Insulin-like growth factor-I (IGF-I) is an essential growth factor that regulates the processes necessary for cell proliferation, differentiation, and survival. The Igf1 gene encodes mature IGF-I and a carboxy-terminal extension called the E-peptide. In rodents, alternative splicing and post-translational processing
Y-F Huang et al.
British journal of cancer, 99(7), 1096-1102 (2008-09-11)
This study was aimed to identify the expression and the correlation of insulin-like growth factor-1 (IGF-1) system and their prognostic impacts in cervical cancer. Seventy-two patients with early-stage cervical cancer were eligible. We obtained the serum levels of total IGF-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service