Skip to Content
Merck
All Photos(1)

Key Documents

I2526

Sigma-Aldrich

Insulin-like Growth Factor-II human

≥98% (SDS-PAGE and HPLC), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

IGF-II

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Insulin-like Growth Factor-II human, IGF-II, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE and HPLC)

form

lyophilized powder

potency

≤2.0 ng/mL ED50

quality

endotoxin tested

mol wt

7.5 kDa

packaging

pkg of 5X50 μg
pkg of 50 μg

technique(s)

cell culture | mammalian: suitable

impurities

≤0.1 EU/μg

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IGF2(3481)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

IGF-II, also known as multiplication stimulating activity (MSA), shows virtually identical bioactivities as IGF-I. IGF-II binds to the IGF-II receptor, IGF-I receptor, and weakly to the insulin receptor (IR). All IGF-II bioactivities in adult tissues are generally attributed to its interaction with IGF-I receptor. However, during embryogenesis the roles of both IGF-II receptor and IR may be significant in promoting normal growth.
Insulin-like growth factor II (IGF-II) also known as multiplication stimulating activity (MSA), shows virtually identical bioactivities as IGF-I. Expression of IGF-II is highest in fetal tissues, where it is believed to play a major role in overall growth and development. After birth IGF-II expression decreases to much lower levels in a variety of tissues. IGF-II is produced also in several tumors. IGF-II binds to IGF-II receptor (IGF-IIR), to IGF-IR and weakly to the insulin receptor (IR). All IGF-II bioactivities in adult tissues are generally attributed to its interaction with IGF-IR. However, during embryogenesis the roles of both IGF-IIR and IR may be significant in promoting normal growth.

Physical form

Lyophilized from a 0.2 μm filtered solution with no additives.

Analysis Note

The biological activity is measured by a cell proliferation assay using mouse FDC-P1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongming Yuan et al.
Scientific reports, 7(1), 78-78 (2017-03-03)
Insulin-like growth factors (IGFs) regulate diverse processes including energy metabolism, cell proliferation and embryonic development. They activate the IGF signaling pathway via binding to cell surface receptors. Here we report an essential role of IGF2 in maintaining the pluripotency of
Şeyda Berk et al.
Frontiers in oncology, 11, 645732-645732 (2021-07-23)
There are only a few experimental studies which have investigated effects of glucose alone, and glucose in combination with insulin/insulin-like growth factors (IGF) on the growth of colon cancer. In the present study, we studied in vitro in human colorectal
Chidambaram Ramanathan et al.
Methods in molecular biology (Clifton, N.J.), 1755, 49-64 (2018-04-20)
In mammals, many aspects of metabolic, physiological, and behavioral processes are regulated by endogenous circadian clocks. Oscillators of different tissue types share a common molecular mechanism at the cellular and molecular level which underlies the rhythmic expression of genes. Individual
Jasmin Svinka et al.
Journal of molecular medicine (Berlin, Germany), 95(1), 109-117 (2016-08-29)
We have demonstrated that the signal transducer and activator of transcription 3 (STAT3) protects from cholestatic liver injury. Specific ablation of STAT3 in hepatocytes and cholangiocytes (STAT3∆hc) aggravated liver damage and fibrosis in the Mdr2-/- (multidrug resistance 2) mouse model
Yi Chen et al.
Molecular oncology, 14(5), 1101-1117 (2020-03-03)
Ewing sarcomas (ESs) are aggressive sarcomas driven by EWS fusion genes. We sought to investigate whether whole-transcriptome sequencing (RNA-seq) could be used to detect patterns associated with chemotherapy response or tumor progression after first-line treatment. Transcriptome sequencing (RNA-seq) of 13

Articles

Insulin-like Growth Factors (IGF)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service