Skip to Content
Merck
All Photos(1)

Key Documents

AP132P

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated

1 mg/mL (after reconstitution), Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

concentration

1 mg/mL (after reconstitution)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

wet ice

target post-translational modification

unmodified

General description

Affinity purified goat anti-rabbit, HRP conjugated.

Specificity

Specific for rabbit IgG, heavy and light chain.

Application

ELISA and Western Blots:
1:5,000-1:100,000 dilution can be used.

Immunohistochemistry:
1:500-1:5,000 dilution can be used.

Optimal working dilutions must be determined by end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
This Goat anti-Rabbit IgG Antibody, Peroxidase Conjugated is validated for use in ELISA, IH, WB for the detection of Rabbit IgG.

Physical form

ImmunoAffinity Purified
Purified goat IgG conjugated to horseradish peroxidase in buffer containing 0.01M PBS,  pH 7.1, with 15mg/mL BSA. Lyophilized.
RECONSTITUTION:
Reconstitute with sterile distilled water to match the volume indicated on the vial label.

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months from date of shipment.
After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.
WARNING: Use of sodium azide as a preservative will substantially inhibit the enzyme activity of HRP.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear NF-kappaB p65 phosphorylation at serine 276 by protein kinase A contributes to the malignant phenotype of head and neck cancer.
Arun, P; Brown, MS; Ehsanian, R; Chen, Z; Van Waes, C
Clinical cancer research : an official journal of the American Association for Cancer Research null
Malihe Aveseh et al.
The Journal of physiology, 593(12), 2635-2648 (2015-04-25)
Monocarboxylate transporters (MCTs) and lactate dehydrogenase A (LDH-A) play important roles in sustaining the glycolytic phenotype seen in cancer. Endurance training improves aerobic capacity; however, whether endurance training alters the metabolic phenotype of a solid tumour, from the perspective of
Harry C Blair et al.
Laboratory investigation; a journal of technical methods and pathology, 96(7), 763-772 (2016-04-19)
Imbalances in lipid metabolism affect bone homeostasis, altering bone mass and quality. A link between bone mass and high-density lipoprotein (HDL) has been proposed. Indeed, it has been recently shown that absence of the HDL receptor scavenger receptor class B
Steven C Pruitt et al.
Nature communications, 4, 2626-2626 (2013-10-24)
Insufficient cell proliferation has been suggested as a potential cause of age-related tissue dysgenesis in mammals. However, genetic manipulation of cell cycle regulators in the germ lines of mice results in changes in animal size but not progeroid phenotypes. Here
Neri Mercatelli et al.
PloS one, 3(12), e4029-e4029 (2008-12-25)
MiR-221 and miR-222 are two highly homologous microRNAs whose upregulation has been recently described in several types of human tumors, for some of which their oncogenic role was explained by the discovery of their target p27, a key cell cycle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service