Skip to Content
Merck
All Photos(1)

Key Documents

AP132

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody

2 mg/mL, Chemicon®

Synonym(s):

Anti-Rabbit IgG, Goat Anti-Rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®

concentration

2 mg/mL

technique(s)

ELISA: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Reacts with Rabbit IgG gamma chain, as well as the light chains from all Rabbit immunoglobulin classes. Monospecific on IEP against whole Rabbit Serum.

Immunogen

Highly purified Rabbit IgG isolated from pooled normal Rabbit Serum.

Application

Goat anti-Rabbit IgG Antibody is an antibody against Rabbit IgG for use in ELISA, IP & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Storage and Stability

Store at +2-8°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kenichiro Sakaguchi et al.
Reproductive biology and endocrinology : RB&E, 17(1), 88-88 (2019-11-07)
The antral follicle count (AFC) in mammalian ovaries positively correlates with female fertility. To clarify the causes of differences in fertility between low and high AFC cows, we investigated follicular growth dynamics and hormone concentrations in plasma, follicular fluid, and
Soah Lee et al.
Biomaterials, 131, 111-120 (2017-04-07)
Engineering 3D human cardiac tissues is of great importance for therapeutic and pharmaceutical applications. As cardiac tissue substitutes, extracellular matrix-derived hydrogels have been widely explored. However, they exhibit premature degradation and their stiffness is often orders of magnitude lower than
Justin V Joseph et al.
Cancer letters, 359(1), 107-116 (2015-01-17)
Glioblastoma (GBM) is the most common brain tumor in adults and the mesenchymal GBM subtype was reported to be the most malignant, presenting severe hypoxia and necrosis. Here, we investigated the possible role of a hypoxic microenvironment for inducing a
Fanghao Hu et al.
Nature methods, 15(3), 194-200 (2018-01-16)
Optical multiplexing has a large impact in photonics, the life sciences and biomedicine. However, current technology is limited by a 'multiplexing ceiling' from existing optical materials. Here we engineered a class of polyyne-based materials for optical supermultiplexing. We achieved 20
Soah Lee et al.
Journal of biomedical materials research. Part A, 104(5), 1064-1071 (2016-01-23)
Smooth muscle tissue is characterized by aligned structures, which is critical for its contractile functions. Smooth muscle injury is common and can be caused by various diseases and degenerative processes, and there remains a strong need to develop effective therapies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service