Skip to Content
Merck
All Photos(1)

Key Documents

20775

Sigma-Aldrich

IHC Select Secondary Goat anti-Mouse IgG, anti-Rabbit IgG, biotinylated

from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405

biological source

goat

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

species reactivity

mouse, rabbit

manufacturer/tradename

Chemicon®
IHC Select

technique(s)

immunohistochemistry: suitable (paraffin)

shipped in

wet ice

target post-translational modification

unmodified

Application

Prediluted for use in Immunohistochemical staining procedures. For example:

Blocking Reagent

Primary Antibody (Mouse IgG or Rabbit IgG) or Negative Control Reagent

Biotinylated Secondary Antibody

Streptavidin-Enzyme Conjugate

Chromogen

Counterstain

For best results use this reagent in conjunction with other IHC Select detection reagents. A complete list of reagents is available at www.chemicon.com.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

Format: Purified
Secondary antibody cocktail containing both Goat Anti-Mouse IgG and Goat Anti-Rabbit IgG, in PBS containing carrier protein, with 0.1% ProClin 300 as preservative.

Storage and Stability

When stored at 2-8°C, this reagent is stable up to the expiration date printed on the label. Do not freeze or expose to elevated temperatures.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Panagiotis Sarantis et al.
International journal of molecular sciences, 22(13) (2021-07-03)
Pancreatic Cancer (PC) is recognized as a highly thrombogenic tumor; thus, low-molecular-weight heparin (LMWH) such as tinzaparin is routinely used for PC patients. On the basis of combinatorial therapy approaches to treat highly malignant and refractory cancers such as PC
Teratoma in the cerebrum of a fantail pigeon.
Celia C Hooper
Avian Pathology : Journal of the W.V.P.A null
Sylvie François et al.
Journal of virology, 90(19), 8686-8697 (2016-07-22)
Varicella-zoster virus (VZV) is an alphaherpesvirus that causes varicella upon primary infection and zoster upon reactivation from latency in sensory ganglion neurons. The replication of herpesviruses requires manipulation of cell signaling pathways. Notably, CREB, a factor involved in the regulation
Li-Wei Dong et al.
Autophagy, 7(10), 1222-1229 (2011-06-10)
Autophagy enables cells to recycle long-lived proteins or damaged organelles. Beclin 1 plays important roles in autophagy, differentiation, apoptosis and the development and progression of cancer, but the expression of Beclin 1 and its possible role in primary intrahepatic cholangiocarcinoma
Riccardo Funari et al.
Biosensors & bioelectronics, 208, 114193-114193 (2022-04-15)
The ongoing emergence of severe acute respiratory syndrome caused by the new coronavirus (SARS-CoV-2) variants requires swift actions in identifying specific antigens and optimizing vaccine development to maximize the humoral response of the patient. Measuring the specificity and the amount

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service