Skip to Content
Merck
All Photos(1)

Key Documents

AP124C

Sigma-Aldrich

Goat Anti-Mouse IgG Antibody, Cy3 conjugate

Chemicon®, from goat

Synonym(s):

Goat IgG Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

CY3 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies
This Goat anti-Mouse IgG Antibody, Cy3 conjugate is validated for use in IF for the detection of Mouse IgG.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junping Yao et al.
Neural regeneration research, 7(16), 1228-1233 (2012-06-05)
Channelrhodopsin-2 ectopically expressed in the retina can recover the response to blue light in genetically blind mice and rats, but is unable to restore visual function due to optic nerve or optic tract lesions. Long Evans rats at postnatal day
Martin Hennenberg et al.
American journal of physiology. Renal physiology, 305(10), F1383-F1390 (2013-09-21)
Inhibition of prostate smooth muscle contraction is an important strategy for medical treatment of lower urinary tract symptoms (LUTS). Besides α1-adrenoceptors, prostate smooth muscle contraction is induced by activation of thromboxane (TXA2) receptors (TXA2-R). Here, we examined the effects of
The roles of fission yeast ase1 in mitotic cell division, meiotic nuclear oscillation, and cytokinesis checkpoint signaling.
Yamashita, A; Sato, M; Fujita, A; Yamamoto, M; Toda, T
Molecular Biology of the Cell null
Telencephalic neurons monosynaptically link brainstem and forebrain premotor networks necessary for song.
Roberts, TF; Klein, ME; Kubke, MF; Wild, JM; Mooney, R
The Journal of Neuroscience null
Akira Ishizuka et al.
Genes to cells : devoted to molecular & cellular mechanisms, 19(9), 704-721 (2014-08-26)
Gomafu/MIAT/Rncr2 is a long noncoding RNA that has been proposed to control retinal cell specification, stem cell differentiation and alternative splicing of schizophrenia-related genes. However, how Gomafu controls these biological processes at the molecular level has remained largely unknown. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service