8.22266
Chromium(VI) oxide
for synthesis
Synonym(s):
Chromium(VI) oxide, Chromic anhydride, Chromium trioxide
About This Item
Recommended Products
Quality Level
form
solid
reaction suitability
reagent type: oxidant
pH
<1 (20 °C, 100 g/L in H2O)
mp
197 °C
density
2.7 g/cm3 at 20 °C
bulk density
900 kg/m3
storage temp.
2-30°C
SMILES string
[Cr](=O)(=O)=O
InChI
1S/Cr.3O
InChI key
WGLPBDUCMAPZCE-UHFFFAOYSA-N
Related Categories
Analysis Note
Identity (ICP-OES): passes test
Signal Word
Danger
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service