Skip to Content
Merck
All Photos(2)

Key Documents

675644

Sigma-Aldrich

Chromium(VI) oxide

99.99% trace metals basis

Synonym(s):

Chromic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CrO3
CAS Number:
Molecular Weight:
99.99
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

powder or flakes

reaction suitability

reagent type: oxidant

mp

196 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromium(VI) oxide (CrO3) is a hexavalent chromium that is majorly used as an oxidizing agent. It oxidizes the C-H bonds in the aromatic rings to form benzoic acid from alkyl benzene. It can be prepared by adding concentrated sulphuric acid in potassium dichromate.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A - Eye Dam. 1 - Muta. 1B - Ox. Sol. 1 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chromium Oxidations in Organic Chemistry (2011)
Evidence for the protective effect of ascorbic acid (vitamin C) in treatment with gamma-rays and chromium (VI) oxide (CrO3) in somatic cells of Drosophila.
O Olvera et al.
Mutation research, 346(1), 19-21 (1995-01-01)
Sara L Holland et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(6), 1764-1767 (2010-07-20)
Toxicity of the environmental carcinogen chromate is known to involve sulfur starvation and also error-prone mRNA translation. Here we reconcile those facts using the yeast model. We demonstrate that: (i) cysteine and methionine starvation mimic Cr-induced translation errors, (ii) genetic
Kyung-Hwa Son et al.
Journal of toxicology and environmental health. Part A, 67(13), 1027-1049 (2004-06-19)
CrO3 is cytotoxic for human epithelial 293 kidney cells over a narrow concentration range of approximately 2-8 microM (D50 approximately 3.0 microM); significantly greater toxicity is observed in clonogenic assays (D50 approximately 0.1-1.0 microM). Survival of a small fraction of
Sethsiri S Samaratunga et al.
Environmental science and pollution research international, 15(1), 27-30 (2008-03-01)
Chromium enters into the aquatic environment as a result of effluent discharge from steel works, electroplating, leather tanning industries and chemical industries. As the Cr(VI) is very harmful to living organisms, it should be quickly removed from the environment when

Articles

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Magnetic materials find diverse applications from data storage to renewable energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service