Skip to Content
Merck
All Photos(1)

Key Documents

359823

Sigma-Aldrich

Chromic acid, polymer-supported

macroporous, 20-50 mesh, extent of labeling: ~2.5 mmol/g loading

Synonym(s):

Chromic acid on Amberlyst® A-26

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

form

macroporous

Quality Level

composition

Carbon content, 43.0-54.0%

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: alcohol reactive

extent of labeling

~2.5 mmol/g loading

particle size

20-50 mesh

SMILES string

O=[Cr](=O)=O

InChI

1S/Cr.3O

InChI key

WGLPBDUCMAPZCE-UHFFFAOYSA-N

Related Categories

Application

Polymer-supported reagent for the clean oxidation of primary and secondary alcohols to carbonyl compounds in high yields.

Legal Information

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Sol. 2 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Cainelli et al.
Journal of the American Chemical Society, 98, 6737-6737 (1976)
Cardillo, G.
Tetrahedron Letters, 3985-3985 (1976)
L.G. Wade et al.
Journal of Chemical Education, 57, 438-438 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service