Skip to Content
Merck
All Photos(2)

Key Documents

8.21202

Sigma-Aldrich

Tris(2-ethylhexyl) phosphate

for synthesis

Synonym(s):

Tris(2-ethylhexyl) phosphate, Phosphoric acid tris(2-ethylhexyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CH2O]3PO
CAS Number:
Molecular Weight:
434.63
MDL number:
UNSPSC Code:
12352108
EC Index Number:
201-116-6
NACRES:
NA.22

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

370 °C

potency

37000 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)

pH

7 (20 °C in H2O)

bp

210 °C/5 hPa (decomposes)

mp

<-70 °C

transition temp

flash point 170 °C

solubility

<0.001 g/L

density

0.92 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[P](=O)(OCC(CCCC)CC)(OCC(CCCC)CC)OCC(CCCC)CC

InChI

1S/C24H51O4P/c1-7-13-16-22(10-4)19-26-29(25,27-20-23(11-5)17-14-8-2)28-21-24(12-6)18-15-9-3/h22-24H,7-21H2,1-6H3

InChI key

GTVWRXDRKAHEAD-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.923 - 0.924
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service