Skip to Content
Merck
All Photos(2)

Key Documents

275794

Sigma-Aldrich

Tris(trimethylsilyl) phosphate

≥98%

Synonym(s):

Phosphoric acid tris(trimethylsilyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3SiO]3P(O)
CAS Number:
Molecular Weight:
314.54
Beilstein:
1794624
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.409 (lit.)

bp

228-229 °C/720 mmHg (lit.)

mp

3-4 °C (lit.)

density

0.945 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)OP(=O)(O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C9H27O4PSi3/c1-15(2,3)11-14(10,12-16(4,5)6)13-17(7,8)9/h1-9H3

InChI key

QJMMCGKXBZVAEI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(trimethylsilyl) phosphate has been investigated as a novel film-forming additive for LiNi0.5Co0.2Mn0.3O2 cycling at high cut-off potential in LiPF6-based electrolyte. Tris(trimethylsilyl) phosphate [Tris(trimethylsilyl) ester of phosphoric acid] undergoes dealkylsilylation reaction with alumazene to yield heteroadamantane molecule.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of [18O4]phosphoric acid in the quantitation of phosphate by gas-liquid chromatography-mass spectrometry analysis.
G Graff et al.
Analytical biochemistry, 107(2), 324-331 (1980-09-15)
Jiri Pinkas et al.
Inorganic chemistry, 41(25), 6914-6918 (2002-12-10)
A dealkylsilylation reaction between alumazene [2,6-(i-Pr)(2)C(6)H(3)NAlMe](3) (1) and tris(trimethylsilyl) ester of phosphoric acid (2) in a 1:3 molar ratio provides the heteroadamantane molecule (MeAl)[2,6-(i-Pr)(2)C(6)H(3)N](3)[Al[OP(OSiMe(3))(3)]](2)(O(3)POSiMe(3)) (3). Compound 3 was characterized by analytical and spectroscopic methods, and its molecular structure was established
Tris (trimethylsilyl) phosphate: A film-forming additive for high voltage cathode material in lithium-ion batteries.
Yan G, et al.
Journal of Power Sources, 248, 1306-1311 (2014)
A Garcia-Casas et al.
Colloids and surfaces. B, Biointerfaces, 181, 973-980 (2019-08-07)
Sol-gel coatings are proposed as surface treatments for titanium-based materials to promote the osseointegration of prosthetic devices with the host. As precursors of sol-gel synthesis, two silanes were selected: 3-methacryloxypropyltrimethoxy silane and 2 tetramethyl orthosilane. Sol-gel synthesis was functionalized with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service