Skip to Content
Merck
All Photos(1)

Key Documents

8.00795

Sigma-Aldrich

1,2-Ethanedithiol

for synthesis

Synonym(s):

1,2-Ethanedithiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6S2
CAS Number:
Molecular Weight:
94.20
MDL number:
UNSPSC Code:
12352105
EC Index Number:
208-752-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

6.4 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

120 mg/kg LD50, oral (Rat)
197 mg/kg LD50, skin (Rabbit)

mp

-41 °C

transition temp

flash point 50 °C

density

1.12 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

SCCS

InChI

1S/C2H6S2/c3-1-2-4/h3-4H,1-2H2

InChI key

VYMPLPIFKRHAAC-UHFFFAOYSA-N

Application

1,2-Ethanedithiol can be used as a reactant to synthesize:
  • Aryl thiols via copper-catalyzed single-step reaction with aryl halides.
  • 1,3-Dithiolanes via Lewis acid-catalyzed chemoselective condensation with carbonyl compounds.
  • Polyalkylene sulfides via polycondensation with dibromides in the presence of NaH.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.123 - 1.126
Identity (IR): passes test

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Fmoc resin cleavage and deprotection are crucial steps for peptide synthesis, yielding the desired peptide after resin detachment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service