Skip to Content
Merck
All Photos(1)

Key Documents

B85404

Sigma-Aldrich

1,4-Butanedithiol

97%

Synonym(s):

1,4-Dimercaptobutane, Tetramethylene dimercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)4SH
CAS Number:
Molecular Weight:
122.25
Beilstein:
1697080
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.529 (lit.)

bp

105-106 °C/30 mmHg (lit.)

density

1.042 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].[O-]S([O-])=O

InChI

1S/C4H10S2/c5-3-1-2-4-6/h5-6H,1-4H2

InChI key

SMTOKHQOVJRXLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Muttenthaler et al.
Nature protocols, 10(7), 1067-1083 (2015-06-19)
Solid-phase peptide synthesis (SPPS) using tert-butyloxycarbonyl (Boc)/benzyl (Bzl) chemistry is an indispensable technique in many laboratories around the globe, and it provides peptides to the pharmaceutical industry and to thousands of scientists working in basic research. The Boc/Bzl strategy has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service