Skip to Content
Merck
All Photos(8)

Key Documents

1.00974

Supelco

Ethanol (denatured)

≥99.5% (GC), reagent grade, for extraction, for analytical purposes, EMSURE®

Synonym(s):

Ethanol denatured, Ethyl alcohol, EtOH, Ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-578-6
NACRES:
NB.21

product name

Ethanol, denatured with about 1% methyl ethyl ketone for analysis EMSURE®

grade

denatured
for analytical purposes
for extraction
reagent grade

Quality Level

vapor pressure

59 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

425 °C (ethanol)

potency

6200 mg/kg LD50, oral (Rat)

expl. lim.

1.3-15 % (v/v) ethanol)

impurities

≤0.00001% Ba (Barium)
≤0.00005% Al (Aluminium)
≤0.00005% B (Boron)
≤0.001% Free acid (as acetic acid)
≤0.1% Water
0.8-1.3% Ethylmethylketone (GC)

evapn. residue

≤0.001%

pH

7 (20 °C)

mp

-114.5 °C (ethanol)

transition temp

flash point 12 °C (ethanol)

density

0.79 g/cm3 at 20 °C

cation traces

Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
K: ≤0.0001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

storage temp.

2-30°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Separation of lignin derivatives from hemp fiber using supercritical CO2, ethanol, and water at different temperatures.: This study demonstrates a method for separating lignin derivatives from hemp fibers using a combination of supercritical CO2, ethanol, and water, highlighting the role of ethanol in enhancing the efficiency of the extraction process, which is crucial for the sustainable production of bio-based materials (Zhang et al., 2024).

  • Elimination of Toxic Solvents from Analytical Methods in Food Analysis: Caffeine Determination in Tea as an Example.: This article discusses the development of analytical methods that eliminate the use of toxic solvents in food analysis, specifically highlighting the replacement of harmful solvents with safer alternatives like ethanol, promoting safer and more environmentally friendly practices (Kalisz et al., 2024).

  • High throughput microfluidics-based synthesis of PEGylated liposomes for precise size control and efficient drug encapsulation.: This study explores the use of ethanol in the high-throughput synthesis of PEGylated liposomes, utilizing microfluidics technology to ensure precise size control and efficient drug encapsulation, which are critical for targeted drug delivery systems (Akar et al., 2024).

  • A novel method for quantitative determination of multiple substances using Raman spectroscopy combined with CWT.: This research introduces a novel approach for the quantitative determination of substances where ethanol is used as a solvent to enhance the spectroscopic analysis, proving crucial for accurate and efficient analytical measurements (Yang et al., 2024).

  • Quality criteria, chemical composition, and antimicrobial activity of the essential oil of Mentha suaveolens Ehrh.: This study involves the extraction of essential oils where ethanol is employed as a solvent to extract active compounds effectively, focusing on the antimicrobial properties and chemical composition of the oils (Afrokh et al., 2024).

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Free acid (as acetic acid): ≤ 0.001 %
Ethylmethylketone (GC): 0.8 - 1.3 %
Readily carbonizable substances: conforms
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.1 %
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.00005 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
K (Potassium): ≤ 0.0001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %

Legal Information

Distribution and use of the following types of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration.
EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

greener alternative product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - (ethanol)

Flash Point(C)

12 °C - (ethanol)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service