1.08323
Toluene
EMPLURA®
Synonym(s):
Toluol, Methylbenzene
About This Item
26 mmHg ( 25 °C)
Recommended Products
vapor density
3.2 (vs air)
Quality Level
vapor pressure
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)
product line
EMPARTA®
Assay
≥99% (GC)
form
liquid
autoignition temp.
997 °F
expl. lim.
7 %
impurities
≤0.002% Free acid (as HCl)
refractive index
n/D 1.496 (lit.)
kinematic viscosity
0.7 cSt(20 °C)
bp
110-111 °C (lit.)
mp
-93 °C (lit.)
transition temp
flash point 4 °C
solubility
0.52 g/L
density
0.865 g/mL at 25 °C (lit.)
storage temp.
2-30°C
SMILES string
Cc1ccccc1
InChI
1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3
InChI key
YXFVVABEGXRONW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Metal Extraction and Digestion: Nitric acid (60%) is extensively used for the extraction and digestion of metals from various samples. This application is crucial for preparing samples for subsequent analysis by techniques such as Inductively Coupled Plasma Mass Spectrometry (ICP-MS) and Atomic Absorption Spectroscopy (AAS). Recent studies have compared the efficiency of nitric acid treatment with microwave digestion for extracting metals from bioprocess samples, demonstrating its effectiveness in achieving high recovery rates and accuracy (Mohammad and Madhavarao, Analytical Biochemistry, 2024).
- Food and Beverage Analysis: Nitric acid is employed in the analysis of food and beverage samples to determine the mineral composition. For instance, the acid is used in microwave-assisted decomposition of foods like rice, carrots, and chayote, facilitating the determination of elements such as aluminum, cadmium, chromium, copper, iron, and lead. This application is vital for ensuring food safety and compliance with health standards (Serafim et al., Brazilian Journal of Analytical Chemistry, 2024).
Packaging
Analysis Note
Identity (IR): conforms
Free acid (as HCl): ≤ 0.002 %
Density (d 20 °C/ 4 °C): 0.867
Readily carbonizable substances: conforms
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.9 °F - closed cup
Flash Point(C)
4.4 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Inorganics and solvents for basic and standard applications manufactured locally.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service