Skip to Content
Merck
All Photos(4)

Key Documents

1.07019

Supelco

Toluene

for analysis EMPARTA® ACS

Synonym(s):

Toluol, Methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-625-9
NACRES:
NB.21
grade:
ACS reagent
bp:
110-111 °C (lit.)
vapor pressure:
22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

grade

ACS reagent

Quality Level

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

product line

EMPARTA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

dilution

(for routine analytical applications)

impurities

≤0.0002 meq/g Acidity
≤0.0006 meq/g Alkalinity
≤0.003% Sulfur compounds (as S)
≤0.03% Water

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n/D 1.496 (lit.)

kinematic viscosity

0.7 cSt(20 °C)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

transition temp

flash point 4 °C

density

0.865 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Experimental and Model-Based Approach to Evaluate Solvent Effects on the Solubility of the Pharmaceutical Artemisinin.: This research investigates the role of toluene as a solvent in enhancing the solubility of artemisinin, using both experimental setups and model-based assessments. The findings offer insights into solvent selection for pharmaceutical synthesis, making significant contributions to solvent theory and practical applications in drug formulation (Wünsche S et al., 2024).

  • Highly sensitive and selective laser-based BTEX sensor for occupational and environmental monitoring.: Describes the development of a new laser-based sensor using toluene as a calibration standard for BTEX (Benzene, Toluene, Ethylbenzene, Xylene) detection. This sensor offers improved sensitivity and selectivity, crucial for monitoring air quality in occupational and environmental health contexts (Mhanna M et al., 2024).

  • Response of microbial communities in aquifers with multiple organic solvent contamination: Implications for MNA remedy.: Explores the ecological impact of toluene and other solvents on microbial communities within aquifers. This study highlights the resilience and adaptability of microorganisms, informing strategies for monitored natural attenuation (MNA) in solvent-contaminated environments (Yang L et al., 2024).

  • Elucidating solvent effects on lipase-catalyzed peroxyacid synthesis through activity-based kinetics and molecular dynamics.: Focuses on the use of toluene to understand its effects on enzyme-catalyzed reactions, specifically lipase-mediated peroxyacid synthesis. The study combines kinetic experiments with molecular dynamics simulations, enhancing the understanding of solvent interactions in biocatalysis (Brandolín SE et al., 2024).

  • Stability-Indicating TLC-Densitometric and HPLC Methods for Simultaneous Determination of Teneligliptin and Pioglitazone in Pharmaceutical Dosage Forms with Eco-Friendly Assessment.: Utilizes toluene in the development of chromatographic methods for the analysis of pharmaceutical compounds. This research demonstrates toluene′s utility in creating efficient, eco-friendly analytical techniques for drug quantification (Akabari AH et al., 2024).

Packaging

1.07019.2521: 2.5L, Minimum order quantity: 4

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0006 meq/g
Boiling point: 109 - 111 °C
Sulfur compounds (as S): ≤ 0.003 %
Readily carbonizable substances: conforms
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.03 %

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service