Skip to Content
Merck
All Photos(2)

Key Documents

1.08327

Supelco

Toluene

for liquid chromatography LiChrosolv®

Synonym(s):

Toluol, Methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
MDL number:
UNSPSC Code:
12191501
EC Index Number:
203-625-9
NACRES:
NA.03

vapor density

3.2 (vs air)

Quality Level

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

product line

LiChrosolv®

grade

isocratic

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

997 °F

expl. lim.

7 %

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0006 meq/g Alkalinity
≤0.05% Water

evapn. residue

≤2.0 mg/L

transmittance

300 nm, ≥70%
310 nm, ≥80%
350 nm, ≥98%

refractive index

n/D 1.496 (lit.)

kinematic viscosity

0.7 cSt(20 °C)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

transition temp

flash point 4 °C

density

0.865 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Influence of toluene on the degradation of complex organic matter in wastewater treatment systems.: This study examines the impact of toluene on the degradation processes of complex organic matter in wastewater treatment systems. It highlights the inhibitory effects of toluene on microbial activity and proposes strategies to mitigate these effects to improve the efficiency of wastewater treatment (Taheri et al., 2022).

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.05 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0006 meq/g
Transmission (at 300 nm): ≥ 70 %
Transmission (at 310 nm): ≥ 80 %
Transmission (from 350 nm): ≥ 98 %
Filtered by 0.2 µm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

Simultaneous analysis of ascorbic acid and dehydroascorbic acid in various food samples by High Performance Thin Layer Chromatography (HPTLC).

Protocols

Silica gel G 254 plates are suitable for analysis of Dextromethorphan following the European pharmacopeia monograph.

Qualitative Thin Layer Chromatography Analysis of Flavonoids and Quantification of Terpene Lactones in Ginkgo Biloba Extracts and Tablets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service