Skip to Content
Merck
All Photos(4)

Key Documents

06-868

Sigma-Aldrich

Anti-GABAA Receptor α1 Antibody

Upstate®, from rabbit

Synonym(s):

Gamma-amino butyric acid receptor subunit alpha-1, GABA(A) receptor subunit alpha-1, gamma-aminobutyric acid (GABA) A receptor, alpha 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

The neurotransmitter GABA (gamma-aminobutyric acid) binds to specific transmembrane receptors in the plasma membrane of both pre- and postsynaptic neurons. Three general classes of GABA receptors are known: GABAA and GABAC (ionotropic) receptors and GABAB (metabotropic) receptors. The GABAA (gamma-Aminobutyric Acid type A) receptor is a multiprotein complex that acts as a ligand-gated chloride channel. These receptors are hetero-oligomers, for which 6 subunits are currently known (a, b, g, d, e and r). There are at least 6 different isoforms of the alpha subunit, and receptor subunit expression varies during development. Increased alpha1 subunit assembly in postsynaptic receptors of cerebellar inhibitory synapses is responsible for the fast inhibitory synaptic currents observed during postnatal development. Additionally, alpha1-containing GABAA receptors are important in regulating the reinforcing properties of alcohol, and expression of the alpha1 subunit increases upon chronic exposure to alcohol, making it a potential therapeutic target for alcohol dependency.

Specificity

Other homologies: Human (94% sequence homology).
This antibody recognizes the topographical domain of GABAA Receptor α1.

Immunogen

Epitope: Topological domain.
KLH-conjugated linear peptide corresponding to the topological domain of rat GABAA Receptor α1.

Application

Immunohistochemistry Analysis: A 1:500 dilution from a representative lot detected GABAA Receptor α1 in rat hippocampus tissue.

Immunoprecipitation Analysis: 5 µL from a representative lot immunoprecipitated GABAA Receptor α1 in rat brain microsomal preparation RIPA lysate.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
This Anti-GABAA Receptor α1 Antibody is validated for use in IH, IP, WB for the detection of GABAA Receptor α1.

Quality

Evaluated by Western Blot in rat brain microsomal preparation tissue lysate.

Western Blot Analysis: A 1:10,000 dilution of this antibody detected GABAA Receptor α1 in 10 µg of rat brain microsomal preparation tissue lysate.

Target description

~51 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat Brain Microsomal Preparation Tissue Lysate.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Segregation of different GABAA receptors to synaptic and extrasynaptic membranes of cerebellar granule cells.
Nusser, Z, et al.
The Journal of Neuroscience, 18, 1693-1703 (1998)
Garrett G Gross et al.
Nature methods, 13(8), 673-678 (2016-06-09)
Although neuronal activity can be modulated using a variety of techniques, there are currently few methods for controlling neuronal connectivity. We introduce a tool (GFE3) that mediates the fast, specific and reversible elimination of inhibitory synaptic inputs onto genetically determined
Shiyong Huang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(48), 15916-15920 (2015-12-04)
Tonic inhibition mediated by extrasynaptic GABA(A) receptors (GABARs) sensing ambient levels of GABA can profoundly alter the membrane input resistance to affect cellular excitability. Therefore, regulation of tonic inhibition is an attractive mechanism to control the levels of cortical firing.
C P Miralles et al.
The Journal of comparative neurology, 413(4), 535-548 (1999-09-25)
A novel anti-beta(3) subunit-specific GABA(A) receptor (GABA(A)R) antibody has been prepared by immunizing a rabbit with a bacterial fusion protein of the large intracellular loop of the beta(3) subunit. The antiserum immunoprecipitated the solubilized GABA(A) receptor. The anti-beta(3) antibody was
Characterization with antibodies of the gamma-aminobutyric acidA/benzodiazepine receptor complex during development of the rat brain.
Vitorica, J, et al.
Journal of Neurochemistry, 54, 187-194 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service