Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501062

Sigma-Aldrich

Anti-GABA-B Receptor antibody produced in rabbit

affinity isolated antibody

Synonym(s):

G protein-coupled receptor 51, GABA-B RECEPTOR, GABABR2, GPR51, R2 SUBUNIT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 108 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GABBR1(2550)

General description

Anti-GABA-B Receptor Antibody detects endogenous levels of total GABA-B Receptor protein.
The γ-aminobutyric acid type B (GABAB) receptor is coded by the GABBR1 gene. It has 22 exons and spans at least 18 kb. GABAB gene is present between an olfactory receptor gene cluster downstream and the HLA (human leukocyte antigen) class I region upstream. It is mapped to human chromosome 6p22.

Immunogen

The antiserum was produced against synthesized peptide derived from human GABA-B Receptor.

Immunogen Range: 871-920

Biochem/physiol Actions

The γ-aminobutyric acid type B (GABAB) is essential for the initiation by agonists like GABA (gamma-aminobutyric acid ). GABAB receptors participates in response habituation. In the visual cortex, GABAB is also involved in the initiation of orientation tuning in conjunction with GABAA receptors.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Association between Single Nucleotide Polymorphisms in Gamma-Aminobutyric Acid B Receptor, Insulin Receptor Substrate-1, and Hypocretin Neuropeptide Precursor Genes and Susceptibility to Obstructive Sleep Apnea Hypopnea Syndrome in a Chinese Han Population.
Li Z, et al.
Medical Principles and Practice : International Journal of the Kuwait University, Health Science Centre, 25(6), 517-524 (2016)
GABBR 1 and SLC 6A1, Two Genes Involved in Modulation of GABA Synaptic Transmission, Influence Risk for Alcoholism: Results from Three Ethnically Diverse Populations.
Enoch MA, et al.
Alcoholism, Clinical and Experimental Research, 40(1), 93-101 (2016)
Different roles for GABAA and GABAB receptors in visual processing in the rat superior colliculus.
Binns KE and Salt TE
The Journal of Physiology, 504(3), 629-639 (1997)
GABBR1 has a HERV-W LTR in its regulatory region?a possible implication for schizophrenia.
Hegyi H.
Biology Direct, 8(1), 5-5 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service