Skip to Content
Merck
All Photos(1)

Key Documents

V-015

Supelco

(−)-Riboflavin (Vitamin B2) solution

100 μg/mL (Methanol:0.1% Ammonium acetate in Water (1:1)), ampule of 1 mL, analytical standard, Cerilliant®

Synonym(s):

(−)-Riboflavin, Vitamin B2, Lactoflavin, Vitamin G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Beilstein:
97825
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard
certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (Methanol:0.1% Ammonium acetate in Water (1:1))

mp

290 °C (dec.) (lit.)

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

CC1=C(C)C=C(N(C[C@H](O)[C@@H]([C@H](O)CO)O)C(C2=N3)=NC(NC2=O)=O)C3=C1

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1

InChI key

AUNGANRZJHBGPY-SCRDCRAPSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

David T C Lin et al.
Journal of refractive surgery (Thorofare, N.J. : 1995), 28(11 Suppl), S841-S848 (2013-03-02)
To report results of a series of highly aberrated corneas treated with a topography-guided excimer laser ablation. Retrospective, nonrandomized, consecutive series of eyes treated with topography-guided photorefractive keratectomy (TG-PRK) with the customized topographical neutralization technique (TNT). Cases included postoperative refractive
Carol P Wilson et al.
Hypertension (Dallas, Tex. : 1979), 61(6), 1302-1308 (2013-04-24)
Intervention with riboflavin was recently shown to produce genotype-specific lowering of blood pressure (BP) in patients with premature cardiovascular disease homozygous for the 677C→T polymorphism (TT genotype) in the gene encoding the enzyme methylenetetrahydrofolate reductase (MTHFR). Whether this effect is
Arundhati Biswas et al.
PloS one, 8(3), e58190-e58190 (2013-03-14)
Two potential orthologs of the human riboflavin transporter 3 (hRFVT3) were identified in the C. elegans genome, Y47D7A.16 and Y47D7A.14, which share 33.7 and 30.5% identity, respectively, with hRFVT3. The genes are tandemly arranged, and we assign them the names
S Keskin et al.
Journal of economic entomology, 106(2), 1058-1063 (2013-06-22)
The objective of this study was to evaluate how storage and Sitophilus granarius L. infestation affects mineral and vitamin (thiamin and riboflavin) contents of wheat grain and flour obtained from the wheat. Wheat samples were infested with nonsexed S. granarius
Ambreen Hafeez et al.
Pakistan journal of pharmaceutical sciences, 26(3), 487-493 (2013-04-30)
Riboflavin (vitamin B2) belongs to a group of respiratory enzymes that occur widely in animals and plants participating in vital oxidation- reduction processes in the body. A computational study was conducted on riboflavin by ArgusLab 4.0.1 to obtain the most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service