Skip to Content
Merck
All Photos(1)

Key Documents

P-090

Supelco

α-Pyrrolidinovalerophenone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO · HCl
CAS Number:
Molecular Weight:
267.79
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=CC=CC=C1)C(CCC)N2CCCC2.Cl

InChI

1S/C15H21NO.ClH/c1-2-8-14(16-11-6-7-12-16)15(17)13-9-4-3-5-10-13;/h3-5,9-10,14H,2,6-8,11-12H2,1H3;1H

InChI key

ROMXVSMENBAYRM-UHFFFAOYSA-N

General description

A certified Snap-N-Spike® solution suitable for use in GC/MS or LC/MS testing methods for bath salt cathinones in clinical toxicology, urine drug testing or forensic analysis applications. Also known as a-PVP or O-2387, a-Pyrrolidinovalerophenone is a derivative of the synthetic cathinones MDPV and pyrovalerone. This illicit drug has stimulant properties and has been found along with MDPV in several bath salt blends across the US, Europe, and Australia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service