Skip to Content
Merck
All Photos(1)

Key Documents

A-109

Supelco

Acetyl fentanyl solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O
CAS Number:
Molecular Weight:
322.44
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

USDEA Schedule I; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C)N(C1CCN(CCC2=CC=CC=C2)CC1)C3=CC=CC=C3

InChI

1S/C21H26N2O/c1-18(24)23(20-10-6-3-7-11-20)21-13-16-22(17-14-21)15-12-19-8-4-2-5-9-19/h2-11,21H,12-17H2,1H3

InChI key

FYIUUQUPOKIKNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Acetyl fentanyl is a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. In June, 2013, the Centers for Disease Control (CDC) issued a health alert for acetyl fentanyl, reporting 14 overdose deaths in Rhode Island.

Application



  • Comparative analysis of fentanyl analogs: A study differentiated structurally similar fentanyl analogs by utilizing Density Functional Theory (DFT) calculations alongside Surface-Enhanced Raman Spectroscopy (SERS), enhancing the precision of forensic and biochemical analysis of compounds like Acetyl fentanyl (Dogruer Erkok et al., 2024).


  • Quantitative detection in toxicology: Research introduced a quantitative LC-MS/MS method for analyzing Acetyl fentanyl along with other fentanyl-related substances in postmortem blood, crucial for accurate forensic investigations and opioid research (Truver et al., 2024).


  • Drug contamination insights: A study explored the attitudes and experiences related to fentanyl contamination in methamphetamine, using self-reports and urine toxicology to underline the presence of fentanyl analogs such as Acetyl fentanyl, impacting public health responses (Daniulaityte et al., 2023).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service