Skip to Content
Merck
All Photos(1)

Key Documents

C-010

Supelco

Cocaethylene solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Benzoylecgonine ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO4
CAS Number:
Molecular Weight:
317.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1[C@@]2([H])[C@@H](C(OCC)=O)[C@@H](OC(C3=CC=CC=C3)=O)C[C@]1([H])CC2

InChI

1S/C18H23NO4/c1-3-22-18(21)16-14-10-9-13(19(14)2)11-15(16)23-17(20)12-7-5-4-6-8-12/h4-8,13-16H,3,9-11H2,1-2H3/t13-,14+,15-,16+/m0/s1

InChI key

NMPOSNRHZIWLLL-XUWVNRHRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for GC/MS or LC/MS methods in clinical toxicology, forensic analysis, or urine drug testing. Cocaethylene is the ethyl ester of benzoylecgonine. This benzoylecgonine analog is formed during cocaine metabolism when ethanol is present.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao Lu et al.
Forensic science international, 189(1-3), 54-59 (2009-05-22)
A reliable, alternative screening method for detection of cocaine and its metabolites, benzoylecgonine and cocaethylene in urine is demonstrated using solid phase extraction (SPE) coupled with ion mobility spectrometry (IMS). Data analysis with alternating least squares (ALS) is used to
Xavier Joya et al.
Forensic science international, 196(1-3), 38-42 (2010-01-09)
We describe the development and validation of a method for the quantification of drugs of abuse, using gas chromatography-mass spectrometry (GC/MS), in human placenta. Concentration ranges covered were 5-500 ng/g for amphetamine, methamphetamine, MDMA, methadone, cocaine, benzoylecgonine, cocaethylene, morphine, 11-nor-9-carboxy-delta-9-tetrahydrocannabinol
Ellen D Herbst et al.
Experimental and clinical psychopharmacology, 19(2), 95-104 (2011-04-06)
Ethanol alters the hepatic biotransformation of cocaine, resulting in transesterification to a novel active metabolite, cocaethylene. Because of first pass metabolism, oral drug administration might be expected to produce relatively larger concentrations of cocaethylene than would intravenous or smoked administration.
Laura Mercolini et al.
Journal of chromatography. A, 1217(46), 7242-7248 (2010-10-12)
An original HPLC method coupled to spectrofluorimetric detection is presented for the simultaneous analysis in dried blood spots (DBS) of cocaine and two important metabolites, namely benzoylecgonine (its main metabolite) and cocaethylene (the active metabolite formed in the presence of
Robert B Parker et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(2), 317-322 (2009-11-19)
Ethanol decreases the clearance of cocaine by inhibiting the hydrolysis of cocaine to benzoylecgonine and ecgonine methyl ester by carboxylesterases, and there is a large body of literature describing this interaction as it relates to the abuse of cocaine. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service