Skip to Content
Merck
All Photos(1)

Key Documents

C1528

Supelco

Cocaine hydrochloride solution

1.0 mg/mL in methanol, analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · HCl
CAS Number:
Molecular Weight:
339.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

drug control

Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.COC(=O)[C@H]1[C@H](C[C@@H]2CC[C@H]1N2C)OC(=O)c3ccccc3

InChI

1S/C17H21NO4.ClH/c1-18-12-8-9-13(18)15(17(20)21-2)14(10-12)22-16(19)11-6-4-3-5-7-11;/h3-7,12-15H,8-10H2,1-2H3;1H/t12-,13+,14-,15+;/m0./s1

InChI key

PIQVDUKEQYOJNR-VZXSFKIWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cocaine is mostly commonly available as cocaine hydrochloride which is a white to off-white powder. It is taken by intranasal route. In some surgeries it is used as topical anaesthetic drug.

Application

Cocaine hydrochloride solution was used as a calibration standard to determine cocaine in human hair samples using gas chromatography/mass spectrometry (GC/MS). It may be used as a standard in the quantitative analysis of cocaine and its adulterants like lidocaine and livamisole using dragendorff reagent combined with paper spray ionization mass spectrometry.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of cocaine and its adulterants (lidocaine and levamisole) using the Dragendorff reagent allied to paper spray ionization mass spectrometry
Santos.H, et al.
Analytical Methods : Advancing Methods and Applications (2017)
L Skender et al.
Forensic science international, 125(2-3), 120-126 (2002-03-23)
Hair of young subjects (N = 36) suspected for drug abuse was analysed for morphine, codeine, heroin, 6-acetylmorphine, cocaine, methadone, amphetamine, methamphetamine, 3,4-methylenedioxyamphetamine (MDA), 3,4-methylenedioxymethamphetamine (MDMA), and 3,4-methylenedioxyethylamphetamine (MDEA). The analysis of morphine, codeine, heroin, 6-acetylmorphine, cocaine, and methadone in
Elena Martín-García et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(10), 2317-2330 (2014-03-19)
High-frequency intake and high drug-induced seeking are associated with cocaine addiction in both human and animals. However, their relationships and neurobiological underpinnings remain hypothetical. The medial prefrontal cortex (mPFC), basolateral amygdala (BLA), and nucleus accumbens (NAc) have been shown to
Billy T Chen et al.
Nature, 496(7445), 359-362 (2013-04-05)
Loss of control over harmful drug seeking is one of the most intractable aspects of addiction, as human substance abusers continue to pursue drugs despite incurring significant negative consequences. Human studies have suggested that deficits in prefrontal cortical function and
John W Muschamp et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(16), E1648-E1655 (2014-04-08)
Hypocretin (orexin) and dynorphin are neuropeptides with opposing actions on motivated behavior. Orexin is implicated in states of arousal and reward, whereas dynorphin is implicated in depressive-like states. We show that, despite their opposing actions, these peptides are packaged in

Protocols

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Analytical standards for drug analysis including heroin, cocaine, amphetamine, and methylenedioxymethamphetamine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service