Skip to Content
Merck
All Photos(1)

Key Documents

M-920

Supelco

Maprotiline hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24ClN
CAS Number:
Molecular Weight:
313.86
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.CNCCCC12CCC(c3ccccc13)c4ccccc24

InChI

1S/C20H23N.ClH/c1-21-14-6-12-20-13-11-15(16-7-2-4-9-18(16)20)17-8-3-5-10-19(17)20;/h2-5,7-10,15,21H,6,11-14H2,1H3;1H

InChI key

NZDMFGKECODQRY-UHFFFAOYSA-N

Gene Information

human ... SLC6A2(6530)

General description

Maprotiline is a tetracyclic antidepressant marketed under the trade name Ludiomil for treatment of depression.  This Certified Spiking Solution® suitable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lynette Hui-Wen Lee et al.
The international journal of neuropsychopharmacology, 12(7), 953-964 (2009-02-11)
Recently, there has been considerable interest in a possible link between changes in brain polyunsaturated fatty acids, neural membrane phospholipid degradation, serotonergic neurotransmission, and depression. The present study aims to examine effects of antidepressants on lipids in different regions of
N Ivkovic et al.
Journal of oral rehabilitation, 35(6), 424-432 (2008-02-21)
The aim of the study was to evaluate the long-term effects of antidepressive therapy on chronic pain and related disability, and masseter silent period in psychiatric depressive patients with temporomandibular disorders (TMD). The study included hospitalized psychiatric depressive patients on
Murat Pinar et al.
Progress in neuro-psychopharmacology & biological psychiatry, 32(1), 135-139 (2007-09-11)
Agents such as clozapine, olanzapine and mirtazapine frequently trigger an increase in body weight. Though the mechanisms have not been thoroughly clarified, recent studies indicate a role for ghrelin in regulation of appetite and weight gain. We investigated the relation
Marwa A Aboukhatwa et al.
BMC neuroscience, 11, 10-10 (2010-01-29)
Recent studies demonstrate that diverse antidepressant agents increase the cellular production of the nucleolipid CDP-diacylglycerol and its synthetic derivative, phosphatidylinositol, in depression-relevant brain regions. Pharmacological blockade of downstream phosphatidylinositide signaling disrupted the behavioral antidepressant effects in rats. However, the nucleolipid
V Hajhashemi et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 59(12), 1053-1059 (2010-06-25)
To explore the site of action of maprotiline, as an atypical antidepressant, on carrageenan-induced paw edema. Male Wistar rats were used. Firstly, the anti-inflammatory effect of systemic maprotiline (12.5, 25 and 50 mg kg(-1)) was assessed using a paw edema

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service