Skip to Content
Merck
All Photos(1)

Key Documents

860501P

Avanti

C22 Ceramide (d18:1/22:0)

Avanti Research - A Croda Brand 860501P, powder

Synonym(s):

N-behenoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H79NO3
CAS Number:
Molecular Weight:
622.06
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860501P-10mg)
pkg of 1 × 5 mg (860501P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860501P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

General description

C22 Ceramide (d18:1/22:0) or N-behenoyl-D-erythro-sphingosine is a synthetic ceramide containing sphingosine with behenic acid.

Application

C22 Ceramide (d18:1/22:0) has been used:
  • as a standard to measure ceramide concentrations in muscle tissue samples using high-performance liquid chromatography/tandem mass spectrometry (LC/MS2)
  • as a standard for the quantitation of sphingolipid metabolites
  • to study its effects on mitochondrial-to-cytosolic stress response (MCSR) induction

Biochem/physiol Actions

Ceramides plays an important role in regulating cell differentiation, proliferation and apoptosis. It also regulates stress responses and inflammation.

Packaging

5 mL Amber Glass Screw Cap Vial (860501P-10mg)
5 mL Amber Glass Screw Cap Vial (860501P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyun-Eui Kim et al.
Cell, 166(6), 1539-1552 (2016-09-10)
Defects in mitochondrial metabolism have been increasingly linked with age-onset protein-misfolding diseases such as Alzheimer's, Parkinson's, and Huntington's. In response to protein-folding stress, compartment-specific unfolded protein responses (UPRs) within the ER, mitochondria, and cytosol work in parallel to ensure cellular
Xingzhong Zhang et al.
Cell metabolism, 30(5), 937-951 (2019-11-02)
Obesity-induced adipose dysfunction is a major contributor to atherosclerosis. Cold exposure has been reported to affect atherosclerosis through regulation of adipose function, but the mechanism has not been well clarified. Here, adipocyte hypoxia-inducible factor 2α (HIF-2α) was upregulated after mild cold
Dimple Kauhanen et al.
Analytical and bioanalytical chemistry, 408(13), 3475-3483 (2016-02-29)
Monitoring the levels of the ceramides (Cer) d18:1/16:0, Cer d18:1/18:0, Cer d18:1/24:0, and Cer d18:1/24:1 and ratios thereof in human plasma empowers the prediction of fatal outcome of coronary artery disease (CAD). We describe a validated liquid chromatography-tandem mass spectrometry
Mutay Aslan et al.
Nutrition and cancer, 1-13 (2020-04-15)
We aimed to investigate the impact of thymoquinone (TQ), on sphingolipid metabolites, ER stress and apoptotic pathways in MCF-7 and HepG2 cancer cells. Antiproliferative effect was exerted in cancer cells via TQ incubation at different doses and durations. Cell viability
Ebru Afşar et al.
Redox biology, 30, 101430-101430 (2020-01-25)
We aimed to determine sphingolipid levels and examine apoptotic pathways in human retinal pigment epithelial cells (ARPE-19) undergoing endoplasmic reticulum (ER) stress. Cells were treated with tunicamycin (TM) to induce ER stress and tauroursodeoxycholic acid (TUDCA), an ER stress inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service