Merck
  • Lipid Biosynthesis Coordinates a Mitochondrial-to-Cytosolic Stress Response.
Lipid Biosynthesis Coordinates a Mitochondrial-to-Cytosolic Stress Response.
Cell (2016-09-10)
Hyun-Eui Kim, Ana Rodrigues Grant, Milos S Simic, Rebecca A Kohnz, Daniel K Nomura, Jenni Durieux, Celine E Riera, Melissa Sanchez, Erik Kapernick, Suzanne Wolff, Andrew Dillin
ABSTRACT
Defects in mitochondrial metabolism have been increasingly linked with age-onset protein-misfolding diseases such as Alzheimer's, Parkinson's, and Huntington's. In response to protein-folding stress, compartment-specific unfolded protein responses (UPRs) within the ER, mitochondria, and cytosol work in parallel to ensure cellular protein homeostasis. While perturbation of individual compartments can make other compartments more susceptible to protein stress, the cellular conditions that trigger cross-communication between the individual UPRs remain poorly understood. We have uncovered a conserved, robust mechanism linking mitochondrial protein homeostasis and the cytosolic folding environment through changes in lipid homeostasis. Metabolic restructuring caused by mitochondrial stress or small-molecule activators trigger changes in gene expression coordinated uniquely by both the mitochondrial and cytosolic UPRs, protecting the cell from disease-associated proteins. Our data suggest an intricate and unique system of communication between UPRs in response to metabolic changes that could unveil new targets for diseases of protein misfolding.
Product Description
Avanti
C22 Ceramide (d18:1/22:0), Avanti Research - A Croda Brand 860501P, powder
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone B-5-1-2, purified from hybridoma cell culture
Avanti
C20 Ceramide (d18:1/20:0), Avanti Research - A Croda Brand 860520P, powder
Sigma-Aldrich
Nile Red, Technical grade