Skip to Content
Merck
All Photos(2)

Key Documents

850475C

Avanti

18:1-16:0 PC

1-oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1--(9Z-octadecenoyl)-2-hexadecanoyl-sn-glycero-3-phosphocholine; OPPC; PC(18:1(9Z)/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H82NO8P
CAS Number:
Molecular Weight:
760.08
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850475C-25mg)
pkg of 2 × 4 mL (850475C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850475C

concentration

10 mg/mL (850475C-25mg)
25 mg/mL (850475C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCC)COC(=O)CCCCCCC\C=C/CCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H82NO8P/c1-6-8-10-12-14-16-18-20-21-23-24-26-28-30-32-34-41(44)48-38-40(39-50-52(46,47)49-37-36-43(3,4)5)51-42(45)35-33-31-29-27-25-22-19-17-15-13-11-9-7-2/h20-21,40H,6-19,22-39H2,1-5H3/b21-20-/t40-/m1/s1

InChI key

RRVPPYNAZJRZFR-VYOBOKEXSA-N

General description

Phosphatidylcholines (PCs) are mainly located on the outer leaflets of the biological membrane bilayer.

Application

18:1-16:0 PC (1-oleoyl-2-palmitoyl-sn-glycero-3-phosphocholine) may be used:
  • as a lipid standard in ultra-performance liquid chromatography (UPLC) analyses
  • as a component in liposomes to study the cell proliferation of several cell lines
  • in liposomes for cross-linking experiment

Biochem/physiol Actions

Phosphatidylcholine (PC) serves as a major source for eicosanoids.

Packaging

5 mL Clear Glass Sealed Ampule (850475C-200mg)
5 mL Clear Glass Sealed Ampule (850475C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Werlein et al.
Biochimica et biophysica acta, 1848(7), 1599-1608 (2015-04-29)
According to early experiments with natural extracts, phosphatidylcholines (PCs) are widely considered essentially non-toxic. In addition to these physiological mixed-chain PCs, many different synthetic diacyl-PCs are currently available, but they have never been systematically evaluated for any interference with cell
Eun A Ra et al.
Nature communications, 7, 11726-11726 (2016-05-25)
Autophagy is responsible for the bulk degradation of cytosolic constituents and plays an essential role in the intestinal epithelium by controlling beneficial host-bacterial relationships. Atg5 and Atg7 are thought to be critical for autophagy. However, Atg5- or Atg7-deficient cells still
Wei He et al.
Nanomedicine : nanotechnology, biology, and medicine, 24, 102154-102154 (2020-01-27)
In vivo delivery of large RNA molecules has significant implications for novel gene therapy, biologics delivery, and vaccine applications. We have developed cationic nanolipoprotein particles (NLPs) to enhance the complexation and delivery of large self-amplifying mRNAs (replicons) in vivo. NLPs
Katharina Wozny et al.
Analytical and bioanalytical chemistry, 411(4), 915-924 (2018-12-24)
Diacyl glycerophospholipids (GPs) belong to the most abundant lipid species in living organisms and consist of a glycerol backbone with fatty acyl groups in sn-1 and sn-2 and a polar head group in the sn-3 position. Regioisomeric mixed diacyl GPs
Jone Garate et al.
Journal of the American Society for Mass Spectrometry, 31(3), 517-526 (2020-03-05)
Imaging mass spectrometry (IMS) is becoming an essential technique in lipidomics. Still, many questions remain open, precluding it from achieving its full potential. Among them, identification of species directly from the tissue is of paramount importance. However, it is not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service