P1652
D-α-Phosphatidylcholine, dipalmitoyl
≥99%
Synonym(s):
2,3-Dihexadecanoyl-sn-glycero-1-phosphocholine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
semisynthetic
Quality Level
Assay
≥99%
form
powder
functional group
phospholipid
lipid type
phosphoglycerides
shipped in
ambient
storage temp.
−20°C
Other Notes
This is the stereoisomer of phosphatidylcholine which is inactive as a substrate for snake venom phospholipase A2.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service