Skip to Content
Merck
All Photos(3)

Key Documents

840021C

Avanti

Egg PE

Avanti Research - A Croda Brand 840021C

Synonym(s):

L-α-phosphatidylethanolamine (Egg, Chicken)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840021C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840021C

concentration

10 mg/mL (840021C-25mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C23H48NO7P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(26)29-20-22(25)21-31-32(27,28)30-19-18-24/h22,25H,2-21,24H2,1H3,(H,27,28)/t22-/m1/s1

InChI key

BBYWOYAFBUOUFP-JOCHJYFZSA-N

General description

Egg PE is a natural phosphatidylethanolamine (PE).

Application

Egg PE has been used as an antigen to determine the outcome of ELISA for antiphosphatidylethanolmine (aPE). It has also been used in protein-lipid overlay (PLO) assay to evaluate the binding of SEIPIN to phosphatidic acid (PA).

Packaging

5 mL Clear Glass Sealed Ampule (840021C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The outcome of ELISA for antiphosphatidylethanolamine antibodies is dependent on the composition of phosphatidylethanolamine
Hou S, et al.
Journal of Immunological Methods, 440, 27-34 (2017)
Daichi Yamaura et al.
Langmuir : the ACS journal of surfaces and colloids, 34(19), 5615-5622 (2018-04-18)
Artificial bilayer lipid membranes (BLMs) provide well-defined systems for investigating the fundamental properties of membrane proteins, including ion channels, and for screening the effect of drugs that act on them. However, the application of this technique is limited due to
Małgorzata A Przeradzka et al.
Journal of thrombosis and haemostasis : JTH, 18(2), 364-372 (2019-11-02)
The identity of the amino acid regions of factor VIII (FVIII) that contribute to factor IXa (FIXa) and von Willebrand factor (VWF) binding has not been fully resolved. Previously, we observed that replacing the FVIII C1 domain for the one
Romina Zanabria et al.
Journal of food biochemistry, 44(2), e13104-e13104 (2019-12-07)
In this work, the known antiproliferative activity of the untreated milk fat globule membrane (MFGM) against human colon cancer cells was employed to test the hypothesis that the supramolecular structure of the MFGM is of important biological significance. The results
Human SEIPIN binds anionic phospholipids
Yan R, et al.
Developmental Cell, 47(2), 248-256 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service