Skip to Content
Merck
All Photos(2)

Key Documents

700061P

Avanti

24(S)-hydroxycholesterol

Avanti Research - A Croda Brand

Synonym(s):

5-cholestene-3β,24-diol; 24-hydroxycholest-5-en-3-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O2
CAS Number:
Molecular Weight:
402.65
UNSPSC Code:
12352211
NACRES:
NA.25

Pricing and availability is not currently available.

description

cholest-5-ene-3β,24(S)-diol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700061P-1mg)
pkg of 1 × 5 mg (700061P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C27H46O2/c1-17(2)25(29)11-6-18(3)22-9-10-23-21-8-7-19-16-20(28)12-14-26(19,4)24(21)13-15-27(22,23)5/h7,17-18,20-25,28-29H,6,8-16H2,1-5H3/t18-,20+,21+,22-,23+,24+,25+,26+,27-/m1/s1

InChI key

IOWMKBFJCNLRTC-XWXSNNQWSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
700071P700021P700036P
shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

manufacturer/tradename

Avanti Research - A Croda Brand

manufacturer/tradename

Avanti Research - A Croda Brand

manufacturer/tradename

Avanti Research - A Croda Brand

manufacturer/tradename

Avanti Research - A Croda Brand

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

packaging

pkg of 1 × 1 mg (700061P-1mg), pkg of 1 × 5 mg (700061P-5mg)

packaging

pkg of 1 × 1 mg (700071P-1mg)

packaging

pkg of 1 × 1 mg (700021P-1mg), pkg of 1 × 25 mg (700021P-25mg), pkg of 1 × 5 mg (700021P-5mg)

packaging

pkg of 1 × 10 mg (700036P-10mg), pkg of 1 × 5 mg (700036P-5mg)

form

powder

form

powder

form

powder

form

powder

General description

24(S)-hydroxycholesterol (24HC) is synthesized from cholesterol in brain dendrites by the action of enzyme cholesterol 24-hydroxylase (CYP46A1).[1] It is catabolized to bile acids in the liver.[2]

Application

24(S)-hydroxycholesterol has been used as an internal standard:
  • in ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)[3]
  • in liquid chromatography with tandem mass spectrometry (LC-MS/MS) for the quantification of 27-hydroxycholesterol from colorectal tumor samples[4]
  • for quantification of human plasma sterols by liquid chromatography with tandem mass spectrometry (LC MS/MS)[5]

Biochem/physiol Actions

24(S)-hydroxycholesterol (24HC) elevated levels are reported in liver inflammation and fibrosis.[2] It is a N-methyl-D-aspartate receptor (NMDAR) modulator.[1] The levels of 24HC are potential indicators of brain development as well as pathology including Alzheimer′s disease (AD) and multiple sclerosis.[6] Polymorphism in the cholesterol 24-hydroxylase (CYP46A1) gene leads to elevated 24HC levels and toxicity. 24HC is a mediator of apoptosis and necroptosis.[7] Elevated levels of 24HC are reported in liver inflammation and fibrosis.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700061P-1mg)
5 mL Amber Glass Screw Cap Vial (700061P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    24S-hydroxycholesterol and 25-hydroxycholesterol differentially impact hippocampal neuronal survival following oxygen-glucose deprivation
    Sun MY, et al.
    PLoS ONE, 12(3) (2017)
    The acidic pathway of bile acid synthesis: Not just an alternative pathway
    Pandak WM and Kakiyama G
    Liver research (2019)
    A validated liquid chromatography-tandem mass spectrometry method for the quantitative determination of 4beta-hydroxycholesterol in human plasma
    van de Merbel NC, et al.
    Journal of Pharmaceutical and Biomedical Analysis, 55(5), 1089-1095 (2011)
    Diverse functions of 24 (S)-hydroxycholesterol in the brain
    Noguchi N, et al.
    Biochemical and Biophysical Research Communications, 446(3), 692-696 (2014)
    24S-hydroxycholesterol and 25-hydroxycholesterol differentially impact hippocampal neuronal survival following oxygen-glucose deprivation
    Sun MY, et al.
    Testing, 12(3) (2017)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service