Skip to Content
Merck
All Photos(3)

Key Documents

830071P

Avanti

Egg Lyso PC

Avanti Research - A Croda Brand 830071P, powder

Synonym(s):

L-α-lysophosphatidylcholine (Egg, Chicken)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 1 g (830071P-1g)
pkg of 1 × 200 mg (830071P-200mg)
pkg of 1 × 25 mg (830071P-25mg)
pkg of 1 × 500 mg (830071P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 830071P

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](O)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C24H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(27)30-21-23(26)22-32-33(28,29)31-20-19-25(2,3)4/h23,26H,5-22H2,1-4H3/t23-/m1/s1

InChI key

ASWBNKHCZGQVJV-HSZRJFAPSA-N

Application

Egg Lyso PC has been used for the preparation of liposomes. It has also been used as a lipid standard for liquid chromatography-mass spectrometry (LC-MS).

Packaging

20 mL Clear Glass Screw Cap Vial (830071P-1g)
20 mL Clear Glass Screw Cap Vial (830071P-500mg)
5 mL Amber Glass Screw Cap Vial (830071P-200mg)
5 mL Amber Glass Screw Cap Vial (830071P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhanced blood?brain barrier transmigration using a novel transferrin embedded fluorescent magneto-liposome nanoformulation
Ding H, et al.
Nanotechnology, 25(5) (2014)
Amperometric post spike feet reveal most exocytosis is via extended kiss-and-run fusion
Mellander LJ, et al.
Scientific reports, 2 (2012)
Amperometric post spike feet reveal most exocytosis is via extended kiss-and-run fusion
Mellander LJ, et al.
Scientific Reports, 2 (2012)
Padma Iyer et al.
PloS one, 7(8), e42627-e42627 (2012-08-24)
Primary open-angle glaucoma is the second leading cause of blindness in the United States and is commonly associated with elevated intraocular pressure (IOP) resulting from diminished aqueous humor (AH) drainage through the trabecular pathway. Developing effective therapies for increased IOP
A relative quantitative positive/negative ion switching method for untargeted lipidomics via high resolution LC-MS/MS from any biological source
Breitkopf S B, et al.
Metabolomics, 13(3), 30-30 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service