Skip to Content
Merck
All Photos(1)

Key Documents

Z317470

Duran® vacuum desiccators

I.D. × H 150 mm × 239 mm, flat flange

Synonym(s):

247825703

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.

material

borosilicate glass

Joint

joint ST/NS24/29

manufacturer/tradename

DURAN®
Schott

availability

not available in USA and Canada

I.D. × H

150 mm × 239 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Duran® vacuum desiccators are designed for drying moist samples and storage of moisture-sensitive substances.
  • The vessel wall thickness enables its working under a vacuum for extremely long periods.
  • Vacuum tight desiccators are manufactured using borosilicate glass 3.3.

Application

Drying of moist samples and storage of moisture-sensitive substances.

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service