Skip to Content
Merck
All Photos(1)

Key Documents

Z119016

Sigma-Aldrich

Scienceware® vacuum desiccator

flange O.D. × I.D. 230 mm × 197 mm

Synonym(s):

Desiccator, Vacuum Bell Jar, Vacuum Desiccator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
eCl@ss:
32040700
NACRES:
NB.43
Pricing and availability is not currently available.

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F42020-0000

clearance

157 mm

flange O.D. × I.D.

230 mm × 197 mm

plate diam.

190 mm

Looking for similar products? Visit Product Comparison Guide

General description

The Scienceware vacuum desiccator has a unique shape which offers maximum interior clearances for more or larger contents without crowding the benchtop. The thick walls ensure a rugged shatter resistant unit that simply stores a vacuum or stays sealed even if not under vacuum. The polypropylene stopcock (PTFE plug) which is included has a vivid, red handle and a hose connection that accepts 1/4 in. (6.4 mm) I.D. vacuum tubing.

The top section can be lifted off using the large ribbed knob at the top center. A neoprene O-ring offers a secure seal, which does not require grease, but is unaffected if grease is applied. A rigid, perforated polypropylene plate, 1/8 in. (3.2 mm) thick is removable and supports the contents, leaving space underneath for desiccant backup if desired.

  • Small footprint with 13% more interior volume than typical desiccator domes
  • Glass transparent, shatter resistant polycarbonate upper section offers unobstructed viewing of contents
  • Polypropylene lower section has excellent chemical resistance and can be used with incandescent crucibles supported on porcelain plate
  • Polypropylene stopcock with PTFE plug controls airflow to gently release vacuum
  • Holds a vacuum of 29.9 in. (759 mm) Hg for 24 hours at room temperature without need for vacuum grease
  • Not compatible with organic solvents and vapors
  • Parts are autoclavable at 121ºC for 20 minutes at 15 psi. All parts that have vacuum grease must have grease reapplied. (Stopcock)

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebecca Soffe et al.
Small (Weinheim an der Bergstrasse, Germany), 16(34), e2002035-e2002035 (2020-07-24)
"After a certain high level of technical skill is achieved, science and art tend to coalesce in aesthetics, plasticity, and form. The greatest scientists are always artists as well." said Albert Einstein. Currently, photographic images bridge the gap between microfluidic/lab-on-a-chip
Rebecca Soffe et al.
PloS one, 14(6), e0218102-e0218102 (2019-06-07)
Artificial surfaces are routinely used instead of leaves to enable a reductionist approach in phyllosphere microbiology, the study of microorganisms residing on plant leaf surfaces. Commonly used artificial surfaces include, flat surfaces, such as metal and nutrient agar, and microstructured
Akio Suzuki et al.
PloS one, 14(6), e0217944-e0217944 (2019-06-07)
Anti-prion protein (PrP) monoclonal antibody 132, which recognizes mouse PrP amino acids 119-127, enables us to reliably detect abnormal isoform prion protein (PrPSc) in cells or frozen tissue sections by immunofluorescence assay, although treatment with guanidinium salts is a prerequisite.

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Do the Scienceware® brand polypropylene products contain any leachable material?

    1 answer
    1. Although Bel-Art does not specifically test their Scienceware® brand polypropylene products, they have no evidence of any problems with leachable material in the polypropylene.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service