Z273546
Krytox® LVP grease
weight 2 oz (tube)
Synonym(s):
Krytox Lubricant
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Du Pont
weight
2 oz (tube)
General description
Krytox® low vapor pressure (LVP) grease is a non-reactive heat-stable lubricant.
Similiar to Krytox 240 AC, except used as high-vacuum grease. 2 oz. tube. R: 36/37/38 S: 26-36
Application
Krytox® LVP grease has been used to seal the holes in septa during Strecker synthesis. It has also been used to lubricate the O-rings of stainless-steel syringe plunger for basic continuous flow equipment: assembly of a reactor coil.
Features and Benefits
- Performs at low pressures (vapor pressures as low as -1.0 x 10-13 torr at 20 °C (68 °F))
- Suitable for extremely high or low temperatures (temperatures ranging from -15 to 300 °C (5 to 572 °F))
- Compatible even in the presence of hostile chemicals or gases
- Outperforms hydrocarbon, silicone, and chlorofluorocarbon greases
Linkage
For more information, see the Krytox LVP data sheet
Legal Information
Krytox is a registered trademark of E. I. du Pont de Nemours and Company
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service