Skip to Content
Merck
All Photos(1)

Key Documents

Z123072

Krytox® 240AC lubricant

pkg of 2 oz

Synonym(s):

Krytox Lubricant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
15121900
NACRES:
NB.35

packaging

pkg of 2 oz

manufacturer/tradename

Du Pont

General description

Krytox®240AC lubricant is a fluorinated grease and is a multi-purpose lubricant.

Application

Krytox® 240AC lubricant is suitable for use in lubricating ground glassjoints and process valves.

Legal Information

Krytox is a registered trademark of E. I. du Pont de Nemours and Company

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applications of FTIR in the identification of foreign materials for biopharmaceutical clinical manufacturing
Li G, et al.
Vibrational Spectroscopy, 50(1), 152-159 (2009)
Lipophilic chloro-oxo-bis (hydroxamato) vanadium (V) complexes: synthesis methods and structure
Dzyuba V I, et al.
Polyhedron, 180, 114421-114421 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service