BR61610
BRAND® stopcock grease
Synonym(s):
Accessories, Stopcock grease, Stopcocks
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
form
grease
packaging
pack of 1 ea
manufacturer/tradename
BRAND 61610
Related Categories
General description
High vacuum quality, silicone-free for lubricating burette stopcocks.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service