WHA77015500
Whatman® UNIPLATE microplates
48 well, flat bottom, well volume 5 mL, natural polypropylene
Synonym(s):
Z746797, micro plate, multiwell plate, well plates
About This Item
natural polypropylene
Recommended Products
material
flat wells
natural polypropylene
sterility
non-sterile
feature
lid: no
packaging
pkg of 25 ea
manufacturer/tradename
Whatman 7701-5500
Whatman Article No. 28420874 (US reference)
technique(s)
PCR: suitable
size
48 well
well maximum volume
5 mL
well volume
5 mL
working volume
5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Sample storage
- Assay development
- High-throughput screening
- Plasmid miniprep
- ELISA assays
- Luminescence/chemiluminescence
- Cell culture
- Filtrate collection
Features and Benefits
- Widest selection from a single source: Choice of well volumes ranging from 8NA µl to 1NA ml, well densities from 24 to 384 wells with round or ′V′ bottom for maximum recovery.
- Chemical compatibility: Available in chemically resistant polymers capable of withstanding low temperatures for long-term storage. Opaque plates prevent optical cross talk in light emitting assays.
- Conforms to ANSI/SBS microplate standard.
- Guaranteed for use with robotic handlers and centrifuge carriers.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service