Skip to Content
Merck
All Photos(1)

Key Documents

WHA30178793

Whatman® cellulose chromatography papers

17-chr, W × L 25 mm × 220 mm

Synonym(s):

Z753599

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115727
NACRES:
NB.24

material

cellulose membrane
sheets

grade

for chromatography

sterility

non-sterile

feature

17-chr

packaging

pk of 100

manufacturer/tradename

Whatman 3017-8793
Whatman Article No. 28419235 (US reference)

parameter

190 mm/30 min flow rate

W × L

25 mm × 220 mm

suitability

suitable for chromatography

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Whatman Grade 17 Chr Cellulose Chromatography Papers are highly absorbent cellulose papers for preparative paper chromatography, Western blotting & nucleic acid blotting. Thick cellulose chromatography paper with uniform wicking and capillary action.

Manufactured and tested specifically for preparative paper chromatography. Whatman Grade 17 Chr Cellulose Chromatography Papers are pure cellulose. Our cellulose is produced entirely from the highest quality cotton linters, with no additives of any kind. They are available in a wide variety of sizes - in both sheets and rolls.

Features and Benefits

  • Wicking capability and uniformity of capillary action: important in chemical separations
  • Durable: suitable for heavy loadings
  • Flow rate: allows for very high flow rates (190 mm/30 min)
  • Widely used: in both protein and nucleic acid blotting

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susana Campuzano et al.
Applied and environmental microbiology, 75(18), 5972-5980 (2009-07-28)
A Streptococcus mitis genomic DNA fragment carrying the SMT1224 gene encoding a putative beta-galactosidase was identified, cloned, and expressed in Escherichia coli. This gene encodes a protein 2,411 amino acids long with a predicted molecular mass of 268 kDa. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service